Skip to Content
Merck
All Photos(1)

Key Documents

234490

Sigma-Aldrich

PD 153035

An extremely potent, cell-permeamble, reversible, ATP-competitive and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC₅₀ = 25 pM; Ki = 6 pM).

Synonym(s):

PD 153035, AG 1517, 4-[(3-Bromophenyl)amino]-6,7-dimethoxyquinazoline, SU 5271, Compound 32

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14BrN3O2
CAS Number:
Molecular Weight:
360.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

25 pM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1cc(ccc1)Nc2ncnc3c2cc(c(c3)OC)OC

InChI

1S/C16H14BrN3O2/c1-21-14-7-12-13(8-15(14)22-2)18-9-19-16(12)20-11-5-3-4-10(17)6-11/h3-9H,1-2H3,(H,18,19,20)

InChI key

LSPANGZZENHZNJ-UHFFFAOYSA-N

General description

An extremely potent, cell-permeamble, reversible, ATP-competitive and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC50 = 25 pM; Ki = 6 pM). Rapidly suppresses autophosphorylation of EGFR at low nanomolar concentrations in fibroblasts or in human epidermoid carcinoma cells. Also selectively blocks EGF-mediated cellular processes including mitogenesis, early gene expression, and oncogenic transformation. A 10 mM (500 µg/139 µl) solution of PD 153035 (Cat. No. 234491) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR
Product competes with ATP.
Reversible: yes
Target Ki: 6 pM against epidermal growth factor receptor (EGFR)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze at -20°C. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bridges, A.J., et al. 1996. J. Med. Chem. 39, 267.
Fry, D.W., et al. 1994. Science265, 1093.

Legal Information

Sold under license of U.S. Patent 5,457,105 and European Patent 0,566,266.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Fry et al.
Science (New York, N.Y.), 265(5175), 1093-1095 (1994-08-19)
A small molecule called PD 153035 inhibited the epidermal growth factor (EGF) receptor tyrosine kinase with a 5-pM inhibition constant. The inhibitor was specific for the EGF receptor tyrosine kinase and inhibited other purified tyrosine kinases only at micromolar or
A J Bridges et al.
Journal of medicinal chemistry, 39(1), 267-276 (1996-01-05)
4-(3-Bromoanilino)-6,7-dimethoxyquinazoline (32, PD 153035) is a very potent inhibitor (IC50 0.025 nM) of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR), binding competitively at the ATP site. Structure-activity relationships for close analogues of 32 are very steep.
Albrecht Stenzinger et al.
Cells, tissues, organs, 184(2), 76-87 (2007-03-16)
The novel protein PTPIP51 (protein tyrosine phosphatase-interacting protein 51), which has been found to interact with protein tyrosine phosphatases of the PTP1B/TcPTP subfamily, is expressed in all suprabasal layers of human epidermis. Hence, a human keratinocyte cell line (HaCaT) grown
Gaurav Agrahari et al.
The Journal of investigative dermatology, 140(3), 656-665 (2019-08-30)
The expressions of LL-37 and KLK-5 were found to be altered in various dermatoses, including atopic dermatitis, psoriasis, and rosacea. However, the downstream inflammatory effect of LL-37 and KLK-5 is not as well studied. In addition, there is little high-quality
Laleh Abbassi et al.
Nature communications, 12(1), 1438-1438 (2021-03-06)
Germ cells are physically coupled to somatic support cells of the gonad during differentiation, but this coupling must be disrupted when they are mature, freeing them to participate in fertilization. In mammalian females, coupling occurs via specialized filopodia that project

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service