233282
Coagulation Factor X, Human Plasma
Coagulation Factor X, Human Plasma, CAS 9001-29-0, is a native coagulation factor X that is converted to Factor Xa, the enzyme that converts prothrombin to thrombin.
Synonym(s):
Coagulation Factor X, Human Plasma, Antihemophilic Factor C, Stuart-Prower Factor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
description
Merck USA index - 14, 3923
form
liquid
specific activity
≥85 units/mg protein (clotting assay)
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Native coagulation factor X from human plasma. In the coagulation cascade, Factor X is converted to Factor Xa, the enzyme that converts prothrombin to thrombin by either the extrinsic or intrinsic pathway. Sold on the basis of units where one unit equals the amount of Factor X in 1 ml pooled human plasma (100 units = 0.8 mg Factor X).
Native coagulation factor X from human plasma. In the coagulation cascade, Factor X is converted to Factor Xa, the enzyme that converts prothrombin to thrombin by either the extrinsic or intrinsic pathway. Sold on the basis of units where one unit equals the amount of factor X in 1 ml pooled human plasma (100 units = 0.8 mg). In the coagulation cascade, Factor X is converted to Factor Xa, the enzyme which converts prothrombin to thrombin by either the extrinsic or intrinsic pathway.
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
In 100 mM NaCl, 20 mM Tris-HCl, 1 mM benzamidine, pH 7.4.
Preparation Note
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Reconstitution
Following initial thaw, aliquot and freeze (-70°C).
Analysis Note
Clotting activity is determined by comparison to a standard curve produced using normal pooled human plasma
single band by SDS-PAGE
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service