Skip to Content
Merck
All Photos(1)

Key Documents

203350

Sigma-Aldrich

Blasticidin S hydrochloride

from Streptomyces griseochromogenes, ≥98% (HPLC), powder, protein syntheis inhibitor, Calbiochem®

Synonym(s):

Blasticidin S, Hydrochloride, Streptomyces griseochromogenes

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N8O5 · xHCl
CAS Number:
Molecular Weight:
422.44 (free base basis)
MDL number:
UNSPSC Code:
51286700
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Blasticidin S, Hydrochloride, Streptomyces griseochromogenes,

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl.N2(C=CC(=NC2=O)N)[C@@H]1O[C@@H]([C@H](C=C1)NC(=O)C[C@@H](N)CCN(C)\C(=N\[H])\N)C(=O)O

InChI

1S/C17H26N8O5.ClH/c1-24(16(20)21)6-4-9(18)8-12(26)22-10-2-3-13(30-14(10)15(27)28)25-7-5-11(19)23-17(25)29;/h2-3,5,7,9-10,13-14H,4,6,8,18H2,1H3,(H3,20,21)(H,22,26)(H,27,28)(H2,19,23,29);1H/t9-,10-,13+,14-;/m0./s1

InChI key

YQXYQOXRCNEATG-NMQKUDMSSA-N

General description

Nucleoside antibiotic that specifically inhibits protein synthesis in both prokaryotes and eukaryotes. Suitable for selection of cells carrying plasmids conferring blasticidin resistance. Blasticidin resistance is conferred by the blasticidin S deaminase genes (bsr from Bacillus cereus or BSD from Aspergillus terreus), whose products convert blasticidin S to a non-toxic deaminohydroxy derivative.
Nucleoside antibiotic that specifically inhibits protein synthesis in both prokaryotes and eukaryotes. Suitable for use as a dominant selectable marker in conjunction with blasticidin S resistant plasmids. Blasticidin S resistance is conferred by the blasticidin S deaminase gene (bcr), which converts blastisidin S to a nontoxic deaminohydroxy derivative.

Biochem/physiol Actions

Primary Target
protein synthesis

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions (pH ≤7) are stable for up to 1 year at -20°C. Avoid alkaline pH. Do not subject to freeze/thaw cycles; upon thawing any aliquots, discard the unused portion.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kimura, M., et al. 1994. Biochim. Biophys. Acta1219, 653.
Kimura, M., et al. 1994, Mol. Gen. Genet.242, 121.
Kojima, N., et al. 1994. J. Biol. Chem.269, 30451.
Kudo, T., et al. 1994. J. Immunol. Methods177, 17.
Sutoh, K. 1993. Plasmid30, 150.
Izumi, M., et al. 1991. Exp. Cell Res.197, 229.
Yamaguchi, H., et al. 1965. J. Biol. Chem.57, 667.
Takeuchi, S., et al. 1958. J. Antibiot.11, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshinobu Kariya et al.
Communications biology, 4(1), 490-490 (2021-04-23)
Epithelial-mesenchymal transition (EMT) plays a pivotal role for tumor progression. Recent studies have revealed the existence of distinct intermediate states in EMT (partial EMT); however, the mechanisms underlying partial EMT are not fully understood. Here, we demonstrate that αvβ3 integrin
Anna Ballard et al.
The Journal of biological chemistry, 295(19), 6629-6640 (2020-03-14)
Dynamic regulation of the mitochondrial network by mitofusins (MFNs) modulates energy production, cell survival, and many intracellular signaling events, including calcium handling. However, the relative importance of specific mitochondrial functions and their dependence on MFNs vary greatly among cell types.
Carole Luthold et al.
Cells, 10(10) (2021-10-24)
The cochaperone BCL2-associated athanogene 3 (BAG3), in complex with the heat shock protein HSPB8, facilitates mitotic rounding, spindle orientation, and proper abscission of daughter cells. BAG3 and HSPB8 mitotic functions implicate the sequestosome p62/SQSTM1, suggesting a role for protein quality
Amy H Ponsford et al.
Autophagy, 17(6), 1500-1518 (2020-06-10)
Disorders of lysosomal physiology have increasingly been found to underlie the pathology of a rapidly growing cast of neurodevelopmental disorders and sporadic diseases of aging. One cardinal aspect of lysosomal (dys)function is lysosomal acidification in which defects trigger lysosomal stress

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service