Skip to Content
Merck
All Photos(1)

Key Documents

20-198

Sigma-Aldrich

cAMP

cAMP used to activate PKA in vitro.

Synonym(s):

Cyclic Adenosine Monophosphate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41

Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

Used to activate PKA in vitro

Physical form

20mM MOPS, 25mM β-glycerol phosphate, 5mM EGTA, 1mM sodium orthovanadate, and 1mM DTT

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service