Skip to Content
Merck
All Photos(1)

Key Documents

20-160

Sigma-Aldrich

1M Tris-HCl, pH 6.5

1M Tris-HCl, pH 6.5.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

Quality Level

form

liquid

manufacturer/tradename

Upstate®

shipped in

wet ice

Application

For use in Chromatin Immunoprecipitation assays.
Research Category
All

Storage and Stability

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Injection-molded nanocomposites and materials based on wheat gluten.
S-W Cho,M Gallstedt,E Johansson,M S Hedenqvist
International Journal of Biological Macromolecules null
Polychlorinated biphenyl congeners in blood of Wisconsin sport fish consumers.
W Sonzogni,L Maack,T Gibson,D Degenhardt,H Anderson,B Fiore
Archives of Environmental Contamination and Toxicology null
Enzyme-linked immunosorbent assays for the detection of antibody to Crimean-Congo haemorrhagic fever virus in the sera of livestock and wild vertebrates.
F J Burt,R Swanepoel,L E Braack
Epidemiology and Infection null
B L Shneider et al.
The Journal of biological chemistry, 268(10), 6985-6988 (1993-04-05)
Taurocholate efflux was studied in Xenopus laevis oocytes and is consistent with a carrier-mediated process. This carrier can be competitively inhibited and trans stimulated by glycocholate. Transport is also trans stimulated by taurochenodeoxycholate and S-hexylglutathione, but not taurolithocholate or daunomycin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service