1.99002
Buffer solution
(potassium dihydrogen phosphate/di-sodium hydrogen phosphate), traceable to SRM from NIST and PTB pH 7.00 (25°C) Certipur®
Synonym(s):
Potassium dihydrogen phosphate/di-sodium hydrogen phosphate buffer solution
About This Item
Recommended Products
grade
certified reference material
Quality Level
product line
Certipur®
pH
7.0 (25 °C in H2O)
bp
109 °C/1013 hPa
mp
-5 °C
density
1.01 g/cm3 at 20 °C
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
- Ready-to-use solution available in 30 mL sachets
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
- Packaging material subjected to comprehensive stability studies as per ISO Guide 35
Legal Information
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service