Skip to Content
Merck
All Photos(1)

Key Documents

196000

Sigma-Aldrich

Bafilomycin A1

from Streptomyces griseus, ≥90% (HPLC), film, V-ATPase inhibitor, Calbiochem

Synonym(s):

Bafilomycin A1, Streptomyces griseus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H58O9
CAS Number:
Molecular Weight:
622.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Bafilomycin A1, Streptomyces griseus, Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.

Quality Level

Assay

≥90% (HPLC)

form

film

shelf life

3 yr

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 100 μM
ethanol: soluble

storage temp.

−20°C

SMILES string

O1[C@@H]([C@H]([C@@H](C[C@]1(O)[C@H]([C@H](O)[C@@H]([C@H]2OC(=O)\C(=C\C(=C\[C@H]([C@H]([C@H](C\C(=C\C=C\[C@@H]2OC)\C)C)O)C)\C)\OC)C)C)O)C)C(C)C

InChI

1S/C35H58O9/c1-19(2)32-24(7)27(36)18-35(40,44-32)26(9)31(38)25(8)33-28(41-10)14-12-13-20(3)15-22(5)30(37)23(6)16-21(4)17-29(42-11)34(39)43-33/h12-14,16-17,19,22-28,30-33,36-38,40H,15,18H2,1-11H3/b14-12+,20-13+,21-16+,29-17-/t22-,23+,24-,25-,26-,27+,28-,30-,31+,32+,33+,35+/m0/s1

InChI key

XDHNQDDQEHDUTM-JQWOJBOSSA-N

General description

A macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase (V-type; Ki = 500 pM). A valuable tool for distinguishing among different types of ATPases. Blocks lysosomal cholesterol trafficking in macrophages and is known to interfere with pH regulation in brain cells. Exhibits cytotoxic effects on a number of cell lines in a cell viability assay. Reported to selectively inhibit β-secretase, an enzyme involved in the processing of amyloid precursor protein (APP). The InSolution format with a purity of ≥97% by HPLC in 90% DMSO is also available (Cat. No. 508409).
Note: 1 set = 3 x 10 µg

Biochem/physiol Actions

Primary Target
vacuolar type H+ ATPase

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 1 year at -20°C.

Other Notes

Sinha, S., and Lieberburg, I. 1999. Proc. Natl. Acad. Sci. USA96, 11049.
Calvert, C.M., and Sanders, D. 1995. J. Biol. Chem.270, 7272.
Nishihara, T., et al. 1995. Biochem. Biophys. Res. Commun.269, 255.
Crider, B.P., et al. 1994. J. Biol. Chem.269, 17379.
Palokangas, H., et al. 1994. J. Biol. Chem.269, 17577.
Sundquist, K.T., and Marks, S.C., Jr. 1994. J. Bone Miner. Res.9, 1575.
Drose, S., et al. 1993. Biochemistry32, 3902.
Furuchi, T., et al. 1993. J. Biol. Chem.268, 27345.
Bowman, E.J., et al. 1988. Proc. Natl. Acad. Sci. USA85, 7972.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joaquim Bosch-Barrera et al.
Cancers, 13(16) (2021-08-28)
The anti-angiogenic agent nintedanib has been shown to prolong overall and progression-free survival in patients with advanced non-small-cell lung cancer (NSCLC) who progress after first-line platinum-based chemotherapy and second-line immunotherapy. Here, we explored the molecular basis and the clinical benefit
Xiangli Bai et al.
Oxidative medicine and cellular longevity, 2020, 9595036-9595036 (2020-07-21)
Subendothelial retention of apolipoprotein B100-containing lipoprotein, such as low-density lipoprotein (LDL), is the initial step of atherogenesis. Activation of autophagy exhibits beneficial effects for the treatment of atherosclerosis. In our previous study, we demonstrated that hyperglycemia suppressed autophagic degradation of
Yulan Liu et al.
Diabetes & vascular disease research, 19(3), 14791641221102513-14791641221102513 (2022-05-14)
This study aims to determine the role and mechanism of autophagy in endothelial cell dysfunction by glucolipotoxicity. Human umbilical vein endothelial cells (HUVECs) were treated with high glucose and high palmitic acid. The number of autophagosomes was evaluated by monodansylcadaverine
Lynne Chantranupong et al.
eLife, 9 (2020-10-13)
Neurons communicate by the activity-dependent release of small-molecule neurotransmitters packaged into synaptic vesicles (SVs). Although many molecules have been identified as neurotransmitters, technical limitations have precluded a full metabolomic analysis of SV content. Here, we present a workflow to rapidly
Yixuan Fang et al.
Aging cell, 19(10), e13232-e13232 (2020-09-21)
Autophagy suppresses mitochondrial metabolism to preserve hematopoietic stem cells (HSCs) in mice. However, the mechanism by which autophagy regulates hematopoietic aging, in particular in humans, has largely been unexplored. Here, we demonstrate that reduction of autophagy in both hematopoietic cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service