Skip to Content
Merck
All Photos(2)

Key Documents

189826

Sigma-Aldrich

5-Aza-2′-Deoxycytidine

≥98% (HPLC), liquid, DNA methyltransferase inhibitor, Calbiochem

Synonym(s):

InSolution 5-Aza-2′-Deoxycytidine, Decitabine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O4
CAS Number:
Molecular Weight:
228.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

5-Aza-2′-Deoxycytidine, InSolution, ≥98%, DNA methyltransferase inhibitor

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−20°C

InChI

1S/C8H12N4O4/c9-7-10-3-12(8(15)11-7)6-1-4(14)5(2-13)16-6/h3-6,13-14H,1-2H2,(H2,9,11,15)

InChI key

XAUDJQYHKZQPEU-UHFFFAOYSA-N

General description

A cytosine analog that acts as a DNA methyltransferase inhibitor. Restores caspase-8 and caspase-10 mRNA and protein expression as well as TRAIL (Tumor necrosis factor-Related Apoptosis Inducing Ligand) sensitivity in TRAIL-resistant cell lines. Also enhances apoptosis induced by HDAC (Histone Deacetylase) inhibitors. The solid form of this compound (Cat. No. 189825) is also available.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Physical form

A 100 mM (10 mg/438 µl) solution of 5-Aza-2ʹ-Deoxycytidine (Cat. No. 189825) in DMSO.

Other Notes

Eggert, A., et al. 2001. Cancer Res.61, 1314.
Takebayashi, S., et al. 2001. Biochem. Biophys. Res. Commun.288, 921.
Zhu, W.G., et al. 2001. Cancer Res.61, 1327.
Hopkins-Donaldson, S., et al. 2000. Cancer Res.60, 4315.
Haaf, T. 1995. Pharmacol. Ther.65, 19.
Jones, P.A., and Taylor, S.M. 1980. Cell20, 85.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liangsheng Wang et al.
The Plant journal : for cell and molecular biology, 109(6), 1397-1415 (2021-12-18)
RNA-directed DNA methylation (RdDM) helps to defend plants against invasive nucleic acids. In the canonical form of RdDM, 24-nt small interfering RNAs (siRNAs) are produced by DICER-LIKE 3 (DCL3). The siRNAs are loaded onto ARGONAUTE (AGO) proteins leading ultimately to
Nanna Jørgensen et al.
International journal of molecular sciences, 21(12) (2020-06-21)
The potential role of human leukocyte antigen (HLA)-G as a target for new cancer immunotherapy drugs has increased the interest in the analysis of mechanisms by which HLA-G expression is regulated, and how the expression can be manipulated. We characterized
Ling-Tong Gao et al.
Cellular and molecular life sciences : CMLS, 80(3), 58-58 (2023-02-07)
The exposure to an unhealthy environment in utero can lead to the occurrence of cardiovascular diseases in the offspring. Glucocorticoids (GC) are essential for normal development and maturation of fetal organs and is a first-line treatment for pregnant women affected
Chuannan Fan et al.
Signal transduction and targeted therapy, 7(1), 126-126 (2022-04-29)
Ovo-like transcriptional repressor 1 (OVOL1) is a key mediator of epithelial lineage determination and mesenchymal-epithelial transition (MET). The cytokines transforming growth factor-β (TGF-β) and bone morphogenetic proteins (BMP) control the epithelial-mesenchymal plasticity (EMP) of cancer cells, but whether this occurs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service