Skip to Content
Merck

189727

Avidin, Fluorescein Conjugate

This Avidin, Fluorescein Conjugate is validated for use in Immunohistochemistry, Immunfluorescence.

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NA.41
UNSPSC Code:
41116133
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Avidin, Fluorescein Conjugate, This Avidin, Fluorescein Conjugate is validated for use in Immunohistochemistry, Immunfluorescence.

biological source

chicken

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

dilution

(Immunohistochemistry (1:100)
Immunfluorescence (1:100))

shipped in

wet ice

storage temp.

−20°C

Quality Level

Application

Immunohistochemistry (1:100)

Immunfluorescence (1:100)

Disclaimer

Toxicity: Standard Handling (A)

General description

Chicken egg white avidin conjugated to fluorescein. Specific for biotin, biotinylated proteins or antibodies.

Other Notes

Label: Protein ratio: (μg/mg) and flourescein : protein (F/P) molar ratio are determined on a lot specific basis. Variables associated with assay conditions will dictate the proper working dilution.

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 150 mM NaCl, 20 mM phosphate buffer, 1% BSA, pH 7.6.

Preparation Note

Following intitial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service