Skip to Content
Merck
All Photos(3)

Key Documents

17-10054

Sigma-Aldrich

ChIPAb+ Histone H2B - ChIP Validated Antibody and Primer Set

from rabbit

Synonym(s):

H2B, Histone H2B, H2B histone family, member Q, histone 2, H2be, histone cluster 2, H2be

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

rabbit

Quality Level

clone

polyclonal

species reactivity

yeast, chicken, Saccharomyces cerevisiae, Xenopus, human

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Related Categories

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Histone H2B set includes the Histone H2B antibody, a negative control rabbit IgG, and qPCR primers which amplify a 110 bp region of human β-globin promoter. The Histone H2B and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Histone H2B-associated chromatin.
Histone H2B is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2A is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Specificity

Broad species cross-reactivity expected based on sequence homology.
Others not tested.
Recognizes and is specific for Histone H2B, MW ~15 kDa. An additional band at ~23 kDa is detected in some samples, which likely corresponds to ubiquityl-Histone H2B.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 118-126 (CG-AVTKYTSSK) of human Histone H2B, with an N-terminal CG added for conjugation purposes.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 1 µL of Normal Rabbit IgG or 1 µL of Anti-Histone H2B and the Magna ChIP A Kit (Cat. # 17-610).
Successful immunoprecipitation of Histone H2B associated DNA fragments was verified by qPCR using ChIP Primers, human β-globin for a positive, and human GAPDH primers as a negative assay (Please see figures).
Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Western Blot Analysis:
Representative lot data.
Recombinant Xenopus Histone H2B, chicken core histones and acid extract from either sodium butyrate or colcemid treated HeLa cells were resolved by electrophoresis, transferred to nitrocellulose and probed with anti-Histone H2B (1:5,000 dilution).
Proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
The ChIPAb+ Histone H2B set includes the Histone H2B antibody, a negative control rabbit IgG & qPCR primers which amplify a 110 bp region of human β-globin promoter.

Packaging

25 assays per set. Recommended use: ~1 μL of antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 1 µL of Normal Rabbit IgG or 1 µL of Anti-Histone H2B and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of Histone H2B associated DNA fragments was verified by qPCR using ChIP Primers, human β-globin (Please see figures).

Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~15 kDa

Physical form

Anti-Histone H2B (rabbit polyclonal). One vial containing 25 µL of protein A purified rabbit polyclonal in buffer containing 0.02 M phosphate buffer, pH 7.6, 0.25 M NaCl, and 0.1% sodium azide, and 30% glycerol. Store at -20°C.

Normal Rabbit IgG. One vial containing 125 μg of purified rabbit IgG in 125 μL storage buffer. Store at -20°C.

ChIP Primers, β-globin. One vial containing 75 μL of 5 μM of each primer specific for the human β-globin promoter. Store at -20°C.
FOR: AGG ACA GGT ACG GCT GTC ATC
REV: TTT ATG CCC AGC CCT GGC TC
Format: Purified
Protein A purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes negative control normal rabbit IgG antibody and primers specific for human β-globin.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianjiang Lan et al.
Molecular and cellular biology, 35(10), 1777-1787 (2015-03-11)
Spinocerebellar ataxia type 7 (SCA7) is a debilitating neurodegenerative disease caused by expansion of a polyglutamine [poly(Q)] tract in ATXN7, a subunit of the deubiquitinase (DUB) module (DUBm) in the SAGA complex. The effects of ATXN7-poly(Q) on DUB activity are
Jiwon Hwang et al.
mBio, 2(2), e00023-e00011 (2011-03-31)
Elongins B and C are members of complexes that increase the efficiency of transcriptional elongation by RNA polymerase II (RNAPII) and enhance the monoubiquitination of histone H2B, an epigenetic mark of actively transcribed genes. Here we show that, in addition
Feng Li et al.
Scientific reports, 6, 36483-36483 (2016-11-08)
Chronic Hepatitis B Virus (HBV) infection is generally not curable with current anti-viral drugs. Virus rebounds after stopping treatment from the stable HBV covalently-closed-circular DNA (cccDNA). The development of drugs that directly target cccDNA is hampered by the lack of
Guang Yang et al.
Nucleic acids research, 48(6), 3001-3013 (2020-01-23)
Nucleosomal histones are barriers to the DNA repair process particularly at DNA double-strand breaks (DSBs). However, the molecular mechanism by which these histone barriers are removed from the sites of DNA damage remains elusive. Here, we have generated a single
Shaun D McCullough et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(52), 21319-21324 (2012-12-14)
Spinocerebellar ataxia type 7 (SCA7) is an autosomal-dominant neurodegenerative disorder that results from polyglutamine expansion of the ataxin-7 (ATXN7) protein. Remarkably, although mutant ATXN7 is expressed throughout the body, pathology is restricted primarily to the cerebellum and retina. One major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service