Skip to Content
Merck
All Photos(1)

Key Documents

1.70321

Supelco

Gold ICP standard

traceable to SRM from NIST H(AuCl₄) in HCl 7% 1000 mg/l Au Certipur®

Synonym(s):

Gold Calibration Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.8

product line

Certipur®

concentration

1000 mg/L Au in 7% HCl

technique(s)

ICP: suitable

density

1.033 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application


  • Trace metal analysis in biomedical research: Gold ICP standard is used in biomedical research to ensure accurate trace metal quantification, fundamental in studies like those assessing cellular uptake of nanoparticles using single-cell ICP-MS (Cassano et al., 2023).

  • Quality control in clinical chemistry: The application of Gold ICP standard in clinical chemistry facilitates precise calibration and quality control, crucial for developing reliable clinical diagnostic tools and ensuring consistency across laboratories (Theodoropoulos et al., 2024).

  • Enhancement of analytical standards: Utilizing Gold ICP standard enhances the accuracy of ICP-MS analysis in the chemical sciences, aiding significant advancements in signal amplification techniques for improved detection sensitivity (Ettedgui et al., 2024).

Other Notes

Blocked in the US and Canada due to regulatory reasons
For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.

Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.

Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.

Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service