Skip to Content
Merck

14-865

p53-GST (recombinant protein)

Synonym(s):

transformation related protein 53

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Form:
liquid
Assay:
~95% (SDS-PAGE)
Biological source:
human
Mol wt:
Mw 72 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Level

assay

~95% (SDS-PAGE)

form

liquid

shelf life

1 yr

mol wt

Mw 72 kDa

manufacturer/tradename

Upstate®

concentration

0.5 mg/mL

technique(s)

activity assay: suitable

solubility

water: soluble

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... TP53(120407068)

General description

Research area: Apoptosis

N-terminal GST-tagged, recombinant full length human P53 protein, expressed in E. coli. Purified using glutathione agarose chromatography. Product Source: Full-length human p53 protein, expressed in e. coli. p53 protein is encoded by the tumor suppressor gene TP53 gene mapped on chromosome 17.

Biochem/physiol Actions

In the cell, the p53 protein binds to DNA, leading to the activation of another gene that produces the p21 protein. This protein interacts with a cell division-stimulating protein (cdk2), preventing the cell from progressing to the next stage of cell division. Mutation in p53 protein leads to uncontrolled cell division resulting in tumor formation. The p53 protein is suggested to impede the phenotypic and genomic changes linked to cancer development through its intricate interaction with various signaling pathways including cell division, genomic stability maintenance, apoptosis, autophagy, immune response, and tumor microenvironment regulation.

Physical form

Glutathione agarose chromatography
Recombinant protein in buffer containing 20 mM Tris-HCl, pH 8.0, 100 mM KCl, 5mM glutathione, 0.5 mM EDTA, 0.5 mM DTT, 1 mM PMSF containing 20% glycerol.

Preparation Note

1 year at -20°C

Analysis Note

Routinely evaluated by SDS-PAGE and Coomassie Stain

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The p53 tumor suppressor protein
Bethesda MD
Genes & diseases (1998)
p53 signaling in cancer progression and therapy
Marei HE, et al.
Cancer Cell International, 21 (2021)
María Pedrero et al.
Biosensors, 6(4) (2016-11-24)
An amperometric magnetoimmunosensor for the determination of human p53 protein is described in this work using a sandwich configuration involving the covalent immobilization of a specific capture antibody onto activated carboxylic-modified magnetic beads (HOOC-MBs) and incubation of the modified MBs



Global Trade Item Number

SKUGTIN
14-86504053252516108