Skip to Content
Merck
All Photos(2)

Key Documents

1.46683

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Sodium thiosulfate, Histidine, Lecithin, Tween®, ICR plus lockable plate, sterile; irradiated, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

TSA, Soybean Casein Digest Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

ready-to-use

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar + LTHTh 90 mm ICR+ settle plate (lockable) is designed for the determination of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains combination of casein and soy peptones by supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with pyruvate to inactivate residues of hydrogen peroxide. Lecithin, polysorbate 80 (Tween®), histidine and sodium thiosulfate are neutralizers to inactivate the active agents of disinfectants such as Alcohol (70% ethanol or isopropyl alcohol), Aldehyde, Chlorite, Dichloroisocyanurate, Glucoprotamine, Peracetic acid, Phenols, quaternary ammonium compounds and Biguanids. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling without the risk of losing the lid. For aerobic incubation the plates should be locked in the “CLOSED”-position (turn the lid clockwise) and for anaerobic or microaerophilic incubation in the “VENT” position (turn the lid anti-clockwise)

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total aerobic microbial count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Settle plates for isolators and critical cleanrooms

Settle plates for isolators and critical cleanrooms

Settle plates for isolators and critical cleanrooms

Settle plates for isolators and critical cleanrooms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service