Skip to Content
Merck
All Photos(1)

Key Documents

14-584-M

Sigma-Aldrich

PAK4 Protein, active, 10 µg

Active, N-terminal His6 & N-terminal GST-tagged, recombinant, human PAK4 amino acids 295-end, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 61.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... PAK4(10298)

General description

Product Source: N-terminal His6 and N-terminal GST-tagged, recombinant, human PAK4 amino acids 295-end
expressed by baculovirus in Sf21 cells

Application

Active, N-terminal His6 & N-terminal GST-tagged, recombinant, human PAK4 amino acids 295-end, for use in Kinase Assays.
Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: PAK4
Target Sub-Family: STE

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-584M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of MBP (Catalog #13-104)

Physical form

Glutathione agarose affinity chromatography

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service