Skip to Content
Merck

Skip To

1.42002

Calcium chloride dihydrate

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP, FCC, E 509

Synonym(s):

Calcium dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Index Number:
233-140-8
E Number:
E509
UNSPSC Code:
12352302
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, Ph. Eur., USP

reg. compliance

FCC

product line

EMPROVE® EXPERT

form

solid

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

mp

176 °C (dec.) (lit.)

solubility

soluble

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Calcium chloride dihydrate is a mineral salt typically used as buffering agent in pharmaceutical formulations. Beyond, calcium chloride dihydrate is used as a complexing builder, flavor enhancer and stabilizer for food and beverage.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10186717258012022
description

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP, FCC, E 509

description

crystalline, Ph. Eur., JP, USP, FCC, E 509

description

extra pure, for table water, FCC, E 509

description

puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

application(s)

pharma/biopharma processes

application(s)

-

agency

BP, JP, USP, ChP, Ph. Eur.

agency

JP, Ph. Eur., USP

agency

-

agency

-

form

solid

form

crystalline, solid

form

solid

form

powder or crystals

product line

EMPROVE® EXPERT

product line

-

product line

-

product line

-

solubility

soluble

solubility

-

solubility

1280 g/L

solubility

-

Quality Level

500

Quality Level

500

Quality Level

300

Quality Level

200


Still not finding the right product?

Explore all of our products under Calcium chloride dihydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters