Skip to Content
Merck
All Photos(1)

Key Documents

12-342

Sigma-Aldrich

Rabbit Anti-Sheep IgG Antibody, HRP conjugate

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

sheep

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Application

Rabbit anti-Sheep IgG Antibody, HRP conjugate detects level of Sheep IgG & has been published & validated for use in ELISA, WB, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated by western blot

Physical form

0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/ml BSA, and 0.01% gentamicin sulfate

Storage and Stability

Lyophilized: 2 years at 4°C; Rehydrated: 6 months at -20°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donel A Sequea et al.
Mechanisms of ageing and development, 134(1-2), 60-63 (2012-12-19)
Moderate calorie restriction (CR) can improve insulin-stimulated Akt phosphorylation and glucose uptake in muscles from 24 month-old rats, but the specific Akt substrates linking CR-effects on Akt to glucose uptake and other cellular processes are uncertain. We probed CR's influence
Susannah L McKay et al.
PloS one, 6(1), e16077-e16077 (2011-02-02)
There is mounting evidence to suggest that the synthesis of pre-mRNA transcripts and their subsequent splicing are coordinated events. Previous studies have implicated the mammalian spliceosomal U2 snRNP as having a novel role in stimulating transcriptional elongation in vitro through
Naveen Sharma et al.
PloS one, 8(6), e65118-e65118 (2013-06-12)
Calorie restriction (CR) (consuming ~60% of ad libitum, AL, intake) improves whole body insulin sensitivity and enhances insulin-stimulated glucose uptake by isolated skeletal muscles. However, little is known about CR-effects on in vivo glucose uptake and insulin signaling in muscle.
Arvin Bansal et al.
Journal of proteomics, 73(10), 2041-2049 (2010-07-06)
Exercise has been shown to improve function of the left ventricle (LV) following myocardial infarction (MI). The mechanisms to explain this benefit have not been fully delineated, but may involve improved mechanics resulting in unloading effects and increased endothelial nitric
Salah Boudjadi et al.
Cancers, 9(8) (2017-09-22)
Integrins are a family of heterodimeric glycoproteins involved in bidirectional cell signaling that participate in the regulation of cell shape, adhesion, migration, survival and proliferation. The integrin α1β1 is known to be involved in RAS/ERK proliferative pathway activation and plays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service