Skip to Content
Merck
All Photos(1)

Key Documents

1.18307

Supelco

Octane-1-sulfonic acid sodium salt

for ion pair chromatography LiChropur

Synonym(s):

Octane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NaO3S
CAS Number:
Molecular Weight:
216.27
MDL number:
UNSPSC Code:
12352107
EC Index Number:
226-195-4
NACRES:
NB.21

Quality Level

Assay

≥99% (acidimetric)

form

solid

technique(s)

ion pair chromatography: suitable

loss

≤2 loss on drying, 120°C; 4h; vacuum

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

5.5-7.5 (20 °C, 100 g/L in H2O)

mp

>300 °C

bulk density

345 kg/m3

suitability

passes test for identity (IR)

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCCCC

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Octane-1-sulfonic acid sodium salt is a commonly used ion-pair reagent in ion-pair chromatography. ​Adding octane-1-sulfonic acid sodium salt to the mobile phase in chromatography can help enhance chromatographic separation by modifying the polarity gap between analytes.

Application

Octane-1-sulfonicacid sodium salt can be used as a mobile phase component in Reverse-PhaseHigh-Performance Liquid Chromatography (RP-HPLC) to facilitate the separation and quantification of amisulpride in the pharmaceutical tablet formulation.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of chromatographic conditions on the separation of selected alkaloids in RP-HPTLC
Petruczynik A, et al.
Journal of Chromatographic Science, 43(4), 183-194 (2005)
Development and validation of an ion-pair HPLC chromatography for simultaneous determination of lactone and carboxylate forms of SN-38 in nanoparticles
Ebrahimnejad P, et al.
Journal of food and drug analysis, 17(4), 8-8 (2009)
Analytical method development and its validation for estimation of amisulpride by reversed phase high performance liquid chromatography (RP-HPLC)
Gupta P, et al.
International Journal of Research in Pharmaceutical Sciences, 4, 542-549 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service