1.15108
M 17 agar acc. to TERZAGHI
suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
form
medium granules (dehydrated (DCM))
Quality Level
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
application(s)
food and beverages
microbiology
storage temp.
15-25°C
suitability
Lactobacillus spp.
Related Categories
General description
for microbiology
Analysis Note
Appearance (clearness): clear
Appearance (colour): brown
pH-value (25 °C): 7.0 - 7.4
Growth (Streptococcus agalactiae ATCC 13813): good to very good
Growth (Lactococcus lactis spp. cremoris ATCC 19257): good to very good
Growth (Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)): good to very good
Growth (Enterococcus faecalis ATCC 11700): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): good to very good
Growth (Lactobacillus acidophilus ATCC 4356 (WDCM 00098)): fair to very good
Growth (Lactobacillus casei ATCC 393 (WDCM 00100)): fair to good
Growth (Lactobacillus fermentum ATCC 9338): fair to good
Incubation: 48 hrs.; 28 °C; aerobic.
Appearance (colour): brown
pH-value (25 °C): 7.0 - 7.4
Growth (Streptococcus agalactiae ATCC 13813): good to very good
Growth (Lactococcus lactis spp. cremoris ATCC 19257): good to very good
Growth (Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)): good to very good
Growth (Enterococcus faecalis ATCC 11700): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): good to very good
Growth (Lactobacillus acidophilus ATCC 4356 (WDCM 00098)): fair to very good
Growth (Lactobacillus casei ATCC 393 (WDCM 00100)): fair to good
Growth (Lactobacillus fermentum ATCC 9338): fair to good
Incubation: 48 hrs.; 28 °C; aerobic.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service