1.14538
COD solution A
for measuring range 4.0 - 40.0; 10 - 150 and 100 - 1500 mg/l; 0.30 ml per determination Spectroquant®
About This Item
Recommended Products
Quality Level
form
liquid
analyte functional class(es)
chemical oxygen demand (COD)
technique(s)
photometry: suitable
pH
<1 (20 °C in H2O)
density
1.25 g/cm3 at 20 °C
storage temp.
15-25°C
General description
For a safer and easier way of COD determination, we recommend to use our Spectroquant COD cell tests where all reagents are already predosed in the celles in defined volumes and ratios for a specific measurement range.
Analysis Note
Assay (H₂SO₄): 14.0 - 14.9 %
Legal Information
Disclaimer

Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT RE 2
Target Organs
Kidney
Storage Class Code
6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service