Recommended Products
specific analyte(s)
ammonium
Quality Level
compatibility
for use with MQUANT®
detection method
colorimetric
storage temp.
15-25°C
General description
Application
- Evaluation of Water Ammonium Ion Test Kit and Its Feasibility for the Analysis of Protein in Food.: This study assesses a water ammonium ion test kit′s application in food protein analysis, highlighting its potential benefits for ensuring food safety and quality (Wu et al., 2022).
- Technical note: Evaluation of a colorimetric point-of-care test for measuring urine ammonium concentration in periparturient dairy cattle.: Discusses a colorimetric point-of-care test for measuring urine ammonium in dairy cattle, crucial for animal health monitoring and management (Megahed & Constable, 2020).
Legal Information
Disclaimer

Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2
Storage Class Code
6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service