1.12130
Dimidium bromide
for surfactant tests
Synonym(s):
Dimidium bromide, 3,8-Diamino-5-methyl- 6-phenylphenanthridinium bromide
About This Item
Quality Level
form
solid
composition
dye content, ≥95% spectrophotometric assay
loss
≤5% loss on drying, 110°C
mp
242 °C
bulk density
380 kg/m3
λmax
523-528 nm in methanol
storage temp.
2-30°C
SMILES string
Br.N1(C2=CC(=N)C=CC2=C3C(=C1c4ccccc4)C=C(C=C3)N)C
InChI
1S/C20H17N3.BrH/c1-23-19-12-15(22)8-10-17(19)16-9-7-14(21)11-18(16)20(23)13-5-3-2-4-6-13;/h2-12,22H,21H2,1H3;1H
InChI key
MQOKYEROIFEEBH-UHFFFAOYSA-N
Application
- A comparison of the base-pair specificities of three phenanthridine drugs using solution spectroscopy.: This study evaluates the specificity of three phenanthridine drugs, including Dimidium bromide, in binding to DNA base pairs through solution spectroscopy techniques. The research offers significant insights into the interactions at the molecular level, providing valuable information for chemists involved in drug design and DNA interaction studies (Dougherty G, 1982).
Analysis Note
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (methanol): 523 - 528 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.0005 %; methanol): 168 - 177
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service