1.09017
TOC standard solution
according to EN 1484-H3, according to DIN 38409-H3, 1000 mg/L potassium hydrogen phthalate in water (stabilized), Certipur®
Synonym(s):
Total Organic Carbon Standard Solution
About This Item
Recommended Products
Agency
according to DIN 38409-H3
according to EN 1484-H3
Quality Level
product line
Certipur®
concentration
1000 mg/L potassium hydrogen phthalate in water (stabilized)
technique(s)
TOC: suitable
storage temp.
15-25°C
General description
Total organic carbon (TOC) is a measure of the total amount of carbon in an organic compound. It is a critical parameter to evaluate the water quality for possible organic contamination.
Application
Features and Benefits
- Ready-to-use solution
- Traceable to the SI and higher order standards from NIST
- Assigned value based on a purity determination by mass balance and gravimetrically prepared value
- Homogeneity assessed in accordance with ISO Guide 35
- Available in high-density polyethylene bottles of volume 100 mL to assure safe delivery
- Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
- Prepared using deionized water as a solvent
Analysis Note
Measurement uncertainty: ± 10 mg/l
TOC: 990 - 1010 mg/l
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service