Skip to Content
Merck
All Photos(1)

Documents

1.07865

Supelco

Strontium chloride hexahydrate

for analysis EMSURE® ACS

Synonym(s):

Strontium chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Sr · 6H2O
CAS Number:
Molecular Weight:
266.62
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-971-6
NACRES:
NA.21

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

98.0-103.0% (complexometric)

form

solid

potency

1796 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

5-7 (25 °C, 50 g/L in H2O)

mp

61 °C

solubility

2058 g/L

density

1.95 g/cm3 at 20 °C

bulk density

1050 kg/m3

anion traces

nitrate (NO3-): ≤0.002%
sulfate (SO42-): ≤0.001%

cation traces

Ba: ≤0.002%
Ca: ≤0.05%
Cu: ≤0.0002%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0002%
Na: ≤0.01%
Pb: ≤0.0002%
heavy metals: ≤0.0005%

storage temp.

2-30°C

InChI

1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2

InChI key

AMGRXJSJSONEEG-UHFFFAOYSA-L

Analysis Note

Assay (complexometric): 99.0 - 103.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water; 25 °C): 5.0 - 7.0
Nitrate (NO₃): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (ACS): ≤ 0.0005 %
Ba (Barium): ≤ 0.002 %
Ca (Calcium): ≤ 0.05 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.0002 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.0002 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shweta Hegde et al.
Pesticide biochemistry and physiology, 167, 104588-104588 (2020-06-13)
Methyl parathion (MP) is a commonly used organophosphorus insecticide in commercial farming. It is well known that MP exposure can affect the function of nervous, respiratory, cardiovascular and reproductive systems. In our previous report we have demonstrated that MP exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service