Skip to Content
Merck
All Photos(2)

Key Documents

1.07657

Millipore

Lactose monohydrate

milk sugar, suitable for microbiology

Synonym(s):

Lactose monohydrate, Milk sugar, Lactobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-559-2
NACRES:
NA.74

Quality Level

form

solid

autoignition temp.

390 °C

potency

>10000 mg/kg LD50, oral (Rat)

pH

4.0-6.5 (20 °C, 100 g/L in H2O)

mp

202 °C

solubility

161 g/L

bulk density

500 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5+,6+,7-,8-,9-,10-,11+,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-XLOQQCSPSA-N

Analysis Note

Identity (IR-spectrum): passes test
Appearance: White to yellowish, finely cristallized powder
Spec. rotation (α 20/D; 10 %; water; calc. on anhydrous substance): +54.4 - +55.9 °
Acidity or alkalinity: passes test
Loss on drying (80 °C, 2 h): ≤ 0.5 %
Water: 4.5 - 5.5 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service