Skip to Content
Merck

1.07553

Extran® MA 02

greener alternative

liquid, neutral, concentrate

Synonym(s):

TM=["EXTRAN"] MA 02 cleaning agent; TM=["EXTRAN"] MA 02 detergent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
41103206
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

7.5 (20 °C, 50 g/L in H2O)

bp

100 °C

density

1.07 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

General description

EXTRAN® MA 02 is a reliable cleaning agent of consistent composition. It is a greener alternative product that is used to clean appliances made from alkali-sensitive metals such as aluminum, zinc, and alloys with similar behavior. It is also suitable for metal appliances and precision measuring devices made from glass and quartz such as burettes, pipettes, cells, blood gas analyzers, and other medical equipment which are sensitive to aggressive detergents and also rarely have problematic contamination.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Application

EXTRAN® MA 02 can be used as a detergent to clean polyester substrate in an ultrasound bath.

Features and Benefits

  • Reliable results
  • Environment-friendly with biodegradable active ingredients
  • Reliable residue-free cleaning with validation support
  • Non-toxic
  • Highly flexible and safe

Analysis Note

pH-value (5 %; water): 6 - 8
Density (d 20 °C/ 4 °C): 1.057 - 1.067
Foaming volume (5 %; water): ≥ 80 ml

Other Notes

Replaces: EX0994; EX0994-4

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
107553250004022536691671
107553010004022536091921
107553500004027269311476
107553902504022536091969
107553901004022536091952
107553100004065268346686