Skip to Content
Merck
All Photos(2)

Key Documents

1.07210

Supelco

Hydrogen peroxide 30%

(Perhydrol) (stabilized for higher storage temp.) for analysis EMSURE ISO

Synonym(s):

Hydrogen peroxide solution, Perhydrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
NACRES:
NA.21
Pricing and availability is not currently available.

Agency

reag. ISO

Quality Level

vapor pressure

18 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

product line

EMSURE®

form

liquid

concentration

≥30.0%

pH

≤3.5 (20 °C in H2O)

bp

107 °C/1013 hPa

mp

-25.7 °C

solubility

water: miscible

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • A method for the extraction of microplastics from solid biowastes including biosolids, compost, and soil for analysis by µ-FTIR.: This research outlines a method using 30% hydrogen peroxide to extract microplastics from various environmental samples, showcasing its utility in environmental analytical chemistry (Ruffell H et al., 2024).


  • Uncovering microplastics contamination in canned seafood.: This study employs 30% hydrogen peroxide to digest organic matter in seafood samples, facilitating the identification of microplastics through chemical analysis techniques, reflecting its application in food safety and environmental sciences (Silva DM et al., 2024).


  • Ion exchange synthesis of copper-based hydroxyapatite for the catalytic degradation of phenol.: In this study, 30% hydrogen peroxide is used to oxidize phenol compounds in wastewater treatments, illustrating its effectiveness in industrial and environmental applications (Peng Y et al., 2023).


Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany
PERHYDROL is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

1–3 of 3 Questions  
  1. Is this food grade at all? thank you. 

    1 answer
    1. This product is intended for research use only. Currently, a food grade product option is not available.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service