Skip to Content
Merck
All Photos(1)

Key Documents

1.02483

Sigma-Aldrich

Chromium(III) oxide

anhydrous, Technipur®

Synonym(s):

Chromic oxide, Chromium sesquioxide, Chromium(3+) oxide, Chromium(III) trioxide, Dichromium trioxide, Trioxochromium, Chromia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr2O3
CAS Number:
Molecular Weight:
151.99
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.21
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

density

5.22 g/cm3

SMILES string

O=[Cr]O[Cr]=O

InChI

1S/2Cr.3O

InChI key

QDOXWKRWXJOMAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromium(III) oxide (Cr2O3) is an inorganic compound with wide applications in construction materials, heterogeneous catalysis, enamels, green pigments, plastic, and paints. These listed applications are due to its unique properties like high oxidative resistance and melting temperature. Furthermore, a permanent green coloring in cement is achieved by Cr2O3.[1][2]

Application

Chromium(III) oxide can be used:
  • To dope TiO2 nanocrystallites photoanodes applicable in dye-sensitized solar cells.[3]
  • As a bulk mediator in the modification of carbon paste electrodes applicable in nitric oxide sensing.[4]
Used in a solid-state reaction to form the new ternary oxide, CrVMoO7, can be used in selective oxidation.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional pigments from chromium(III) oxide nanoparticles
Sangeetha S, et al.
Dyes and Pigments, 94(3), 548-552 (2012)
Amperometric Nitric Oxide Sensor Based on Carbon Paste Electrode Modified with Chromium (III) Oxide
Berisha LS, et al.
Sensors & Transducers Journal, 184(1), 159-164 (2015)
Physical properties of anatase TiO2 nanocrystallites: based photoanodes doped with Cr2O3
Abouhaswa AS
Optical and Quantum Electronics, 52 (2020)
Original synthesis of chromium (III) oxide nanoparticles
Gibota P and Vidal L
J. Eur. Ceram. Soc., 30(4), 911-915 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service