Skip to Content
Merck
All Photos(2)

Key Documents

1.01516

Supelco

Sulfuric acid

96%, Ultrapur

Synonym(s):

Sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Sulfuric acid 96%, Ultrapur

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 300

vapor density

<0.3 (25 °C, vs air)

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

Assay

96% (acidimetric)

form

liquid

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

cation traces

Ag: ≤50 ppt
Al: ≤200 ppt
As: ≤20 ppt
Au: ≤20 ppt
Ba: ≤200 ppt
Be: ≤20 ppt
Bi: ≤20 ppt
Ca: ≤1000 ppt
Cd: ≤20 ppt
Ce: ≤20 ppt
Co: ≤20 ppt
Cr: ≤200 ppt
Cu: ≤50 ppt
Dy: ≤20 ppt
Er: ≤20 ppt
Eu: ≤20 ppt
Fe: ≤500 ppt
Ga: ≤20 ppt
Gd: ≤20 ppt
Ge: ≤20 ppt
Hf: ≤10 ppt
Hg: ≤500 ppt
Ho: ≤20 ppt
In: ≤20 ppt
K: ≤500 ppt
La: ≤20 ppt
Li: ≤20 ppt
Lu: ≤20 ppt
Mg: ≤500 ppt
Mn: ≤20 ppt
Mo: ≤50 ppt
Na: ≤500 ppt
Nb: ≤20 ppt
Nd: ≤20 ppt
Ni: ≤100 ppt
Pb: ≤100 ppt
Pd: ≤50 ppt
Pr: ≤20 ppt
Pt: ≤20 ppt
Rb: ≤20 ppt
Sb: ≤20 ppt
Sc: ≤20 ppt
Se: ≤500 ppt
Sm: ≤20 ppt
Sn: ≤20 ppt
Sr: ≤20 ppt
Ta: ≤50 ppt
Tb: ≤20 ppt
Te: ≤50 ppt
Th: ≤10 ppt
Ti: ≤50 ppt
Tl: ≤20 ppt
Tm: ≤20.0 ppt
U: ≤10.00 ppt
V: ≤20 ppt
W: ≤100 ppt
Y: ≤10.0 ppt
Yb: ≤20.0 ppt
Zn: ≤200 ppt
Zr: ≤20.0 ppt

SMILES string

OS(O)(=O)=O

storage temp.

2-30°C

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • A novel premixing strategy for highly sensitive detection of nitrite on paper-based analytical devices.: This study introduces a new premixing method to enhance the sensitivity of nitrite detection using N-(1-naphthyl) ethylenediamine dihydrochloride. The method significantly improves the detection limits and accuracy, making it a valuable tool for various analytical applications (Xu et al., 2024).


  • Electrochemical Self-Assembled Gold Nanoparticle SERS Substrate Coupled with Diazotization for Sensitive Detection of Nitrite.: The study demonstrates the use of N-(1-naphthyl) ethylenediamine dihydrochloride to generate azo compounds, that assemble on the surface of AuNP/ITO an electrochemically self-assembled gold nanoparticle substrate for surface-enhanced Raman spectroscopy (SERS), coupled with diazotization, to detect nitrite with high sensitivity (Han et al., 2022).




Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service