Skip to Content
Merck

1.01136

Ammonium carbonate

EMPROVE® ESSENTIAL, NF

Synonym(s):

Ammonium carbonate, salt of hartshorn

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352301
E Number:
E503
EC Index Number:
233-786-0
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

NF

Quality Level

vapor pressure

69 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

solid

potency

1800-2150 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

pH

9.4 (20 °C, 100 g/L in H2O)

solubility

320 g/L

application(s)

pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[N+H4].N.[O-]C(=O)O

InChI

1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3

InChI key

PRKQVKDSMLBJBJ-UHFFFAOYSA-N

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ammonium carbonate is mostly used as acidic regulator. Ammonium carbonate Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ammonium carbonate


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

wgk

WGK 1



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
101136000004022536016535
101136905104022536016580