Skip to Content
Merck

Skip To

1.00975

n-Amyl alcohol

(Pentan-1-ol) for analysis EMSURE®

Synonym(s):

n-Amyl alcohol, 1-Pentanol, Pentyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
12191502
EC Index Number:
200-752-1
NACRES:
NB.21
MDL number:
Assay:
≥98.5% (GC)
Grade:
reagent grade
Bp:
138 °C/1013 hPa
Vapor pressure:
2.04 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

Quality Segment

vapor pressure

2.04 hPa ( 20 °C)

product line

EMSURE®

assay

≥98.5% (GC)

form

liquid

autoignition temp.

300 °C (DIN 51794)

potency

3670 mg/kg LD50, oral (Rat), 2306 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6 % (v/v)

dilution

(for analytical testing)

impurities

≤0.000002% B (Boron), ≤0.000002% Co (Cobalt), ≤0.000002% Cr (Chromium), ≤0.000005% Cd (Cadmium), ≤0.00001% Ba (Barium), ≤0.00005% Al (Aluminium), ≤0.00005% Ca (Calcium), ≤0.005% Free acid (as CH3COOH), ≤0.1% Valerian aldehyde (GC), ≤0.1% Water

evapn. residue

≤0.005%

pH

7 (20 °C, 22 g/L)

bp

138 °C/1013 hPa

mp

-78 °C

transition temp

flash point 47 °C

density

0.815 g/cm3 at 20 °C

cation traces

Cu: ≤0.000002%, Fe: ≤0.00005%, Mg: ≤0.00001%, Mn: ≤0.000002%, Ni: ≤0.000002%, Pb: ≤0.00001%, Sn: ≤0.00001%, Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

OCCCCC

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Synthesis and Properties of 1,11,15,25-Tetrahydroxy-4,8,18,22-Di (Bridged Dipropionate Carboxyl) Phthalocyanine Copper: This study highlights the application of n-amyl alcohol in the synthesis of complex organic compounds, demonstrating its effectiveness in facilitating high-precision chemical reactions essential for the development of novel materials and pharmaceuticals (Xia et al., 2015).

  • Cr(III)/Cr(VI) speciation determination of chromium in water samples by luminescence quenching of quercetin: This study utilizes n-amyl alcohol for environmental monitoring, specifically for the detection and differentiation of chromium species in water, illustrating its importance in environmental safety and compliance (Hosseini and Belador, 2009).

  • Application of headspace solvent microextraction to the analysis of mononitrotoluenes in waste water samples: The use of n-amyl alcohol in sample preparation for pollutant analysis in wastewater highlights its critical role in environmental testing protocols, providing effective and efficient extraction methods (Ebrahimzadeh et al., 2007).

  • Solvothermal preparation and gas sensing properties of ZnO whiskers: This article discusses the use of n-amyl alcohol in the solvothermal synthesis of zinc oxide whiskers, underlining its utility in the fabrication of sensors that are crucial for environmental monitoring and material science research (Jiaqiang et al., 2006).

Other Notes

Discover EMSURE - to be more than sure!
Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.07500W205605
assay

≥98.5% (GC)

assay

≥98% (GC)

assay

≥99%

Quality Level

300

Quality Level

200

Quality Level

300

grade

reagent grade

grade

synthesis grade

grade

FG, Fragrance grade

form

liquid

form

liquid

form

liquid

expl. lim.

1.6 % (v/v)

expl. lim.

1.6 % (v/v)

expl. lim.

10 %, 100 °F

autoignition temp.

300 °C (DIN 51794)

autoignition temp.

300 °C (DIN 51794)

autoignition temp.

572 °F


Still not finding the right product?

Explore all of our products under n-Amyl alcohol


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters