Skip to Content
Merck

Skip To

1.00784

Rectangular cells quartz 10 mm

Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
SE.11
UNSPSC Code:
41115311
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

quartz vial

product line

Spectroquant®

technique(s)

photometry: suitable

detection method

photometric

storage temp.

2-30°C

General description

Cuvettes made of quartz glas are especially suitable to perform measurements using a wavelength from the UV range. In contrast to other cuvettes, these cells don´t absorb in this range and therefore allow for safe measurement results. Thy can be used in combination with photometes with a cell holder for 10 mm rectangular cells, e.g. the Spectroquant Prove series.
Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.14946Z2766691.14947
material

quartz vial

material

optical glass vial

material

PTFE stopper, quartz

material

optical glass vial

product line

Spectroquant®

product line

-

product line

-

product line

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

detection method

photometric

detection method

photometric

detection method

-

detection method

photometric

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

-

technique(s)

photometry: suitable

microbialanalysis
microbialanalysis

microbialanalysis
microbialanalysis

-

microbialanalysis
microbialanalysis


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters