Skip to Content
Merck

1.00718

BOD Standard

EN 5815-1 (analogous), 198 ± 40 mg/L BOD5, pkg of 10 × 1 L, standard solution Spectroquant®

Synonym(s):

Biochemical oxygen demand standard solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

EN 5815-1 (analogous)

Quality Segment

product line

Spectroquant®

analyte functional class(es)

biochemical oxygen demand (BOD)

packaging

pkg of 10 × 1 L

concentration

198 ±40 mg/L (BOD5)

application(s)

aquaculture
disposal drainage water
drinking water
electroplating
environmental
ground and surface water
seawater

format

single component solution

storage temp.

15-25°C

General description

This lyophilized Spectroquant® BOD Standard is diluted with distilled water to prepare a standard solution of an appropriate concentration.

This standard solution is then analyzed using the Spectroquant® BOD Cell Test to check the procedure for the BOD measurement. The quantity of lyophilisate contained in each bottle of BOD Standard is sufficient to prepare 1 l of standard solution with 198 ± 40 mg/l BOD5 or 206 ± 40 mg/l BOD7.

These Spectroquant® standards perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.

Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.

Application

BOD Standard can be used to check the photometric measurement system (reagents, measurement device, handling) and the mode of working.

Features and Benefits

  • The standard is analogous to EN 5815-1
  • Accompanied with a certificate of analysis (COA)
  • Reliable and reproducible results

Analysis Note

Assay (BSB): 190 - 230 mg/l

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100718000104022536751368