Skip to Content
Merck

Skip To

1.00016

Acetonitrile

≥99.9% (GC), suitable for UV/Vis spectroscopy, Uvasol®

Synonym(s):

Methyl cyanide, Anhydrous ACN, Dry MeCN

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
NACRES:
NA.03
EC Index Number:
200-835-2
UNSPSC Code:
12191502
MDL number:
Beilstein/REAXYS Number:
741857
Assay:
≥99.9% (GC)
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
81-82 °C (lit.)
Vapor pressure:
72.8 mmHg ( 20 °C), 97 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetonitrile, for spectroscopy Uvasol®

Quality Segment

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C), 97 hPa ( 20 °C)

assay

≥99.9% (GC)

form

liquid

autoignition temp.

524 °C, 973 °F

expl. lim.

16 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0001 meq/g Alkalinity, ≤0.0002 meq/g Acidity, ≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

190 nm, ≥20%, 195 nm, ≥60%, 200 nm, ≥90%, 215 nm, ≥95%, 230 nm, ≥98%

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

UV absorption

λ: 190 nm Amax: ≤0.70, λ: 195 nm Amax: ≤0.22, λ: 200 nm Amax: ≤0.05, λ: 215 nm Amax: ≤0.02, λ: 230 nm Amax: ≤0.01

format

neat

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application



  • Anthocyanin Separation: Acetonitrile is employed in high-performance liquid chromatography (HPLC) for the selective separation of anthocyanins. Studies have demonstrated the benefits of replacing acetonitrile with methanol in the mobile phase to control the selectivity and improve the resolution of anthocyanin separation, providing a more sustainable approach to chromatographic analysis (Deineka et al., Journal of Analytical Chemistry, 2021).

  • Detection of α-Amino Acids: Acetonitrile is used in the development of chemiluminescence methods for detecting α-amino acids. A mixed solution of water, acetonitrile, and ethyl acetate has been shown to enhance the sensitivity and accuracy of amino acid detection, making it a valuable tool for biochemical and clinical analyses (Kan et al., American Journal of Analytical Chemistry, 2020).

  • Electrochemical Detection of Biomarkers: Acetonitrile is utilized in the electrochemical detection of biomarkers such as 5-formyluracil. Labeling with (2-benzimidazolyl) acetonitrile enhances the selectivity and sensitivity of the detection, making it an important method for biomedical research and diagnostics (Tang et al., Analytical Chemistry, 2021).

  • Ligand-Assisted Excitation of Europium Complexes: Acetonitrile is used in studying the ligand-assisted excitation of Eu(III) complexes. The interaction of europium complexes with xenon difluoride in acetonitrile solution provides insights into their photophysical properties, which are essential for developing luminescent materials for various applications, including lighting and display technologies (Masyagutova et al., Journal of Fluorine Chemistry, 2024).


Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9006671.000171.13358
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

HPLC: suitable

assay

≥99.9% (GC)

assay

≥99.92%

assay

≥99.8% (GC)

assay

≥99.8% (GC)

form

liquid

form

liquid

form

liquid

form

liquid

UV absorption

λ: 190 nm Amax: ≤0.70, λ: 200 nm Amax: ≤0.05, λ: 230 nm Amax: ≤0.01, λ: 195 nm Amax: ≤0.22, λ: 215 nm Amax: ≤0.02

UV absorption

-

UV absorption

-

UV absorption

-

bp

81-82 °C (lit.)

bp

81-82 °C (lit.)

bp

81-82 °C (lit.)

bp

81-82 °C (lit.)


Still not finding the right product?

Explore all of our products under Acetonitrile


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters