Skip to Content
Merck
All Photos(2)

Key Documents

09-817

Sigma-Aldrich

Anti-JMJD1C Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

JMJD, jumonji domain containing 1C, TR-interacting protein 8, thyroid receptor interacting protein 8, probable JmjC domain-containing histone demethylation protein 2C, thyroid hormone receptor interactor 8, Thyroid receptor-interacting protein 8, Jumonji

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
5

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), orangutan (based on 100% sequence homology), rabbit (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JMJD1C(221037)

General description

JMJD1C (jumonji domain-containing protein 1C) is also known as TRIP8 and mutations in this histone demethylase have been linked to certain forms of autism. JMJD1C is also thought to work in reactivating genes in embryonic stem cells, pancreatic islet and gastric cancer. JMJD1C has also been implicated as a gene expression regulator. JMJD1C with WHISTLE have recently been shown to also play a role in mouse testis development. JMJD1C has been identified as a nuclear-receptor coactivator and is a candidate gene for autism.

Immunogen

KLH-conjugated linear peptide corresponding to human JMJD1C.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected JMJD1C in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Use Anti-JMJD1C Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect JMJD1C also known as jumonji domain containing 1C or TR-interacting protein 8.

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 1 µg/mL of this antibody detected JMJD1C on 10 µg of HeLa nuclear extract.

Target description

~284 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim Verhagen et al.
ACS nano, 14(11), 15587-15594 (2020-10-30)
Various properties of water are affected by confinement as the space-filling of the water molecules is very different from bulk water. In our study, we challenged the creation of a stable system in which water molecules are permanently locked in
Mauricio A Sáez et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 18(4), 378-385 (2015-07-17)
Autism spectrum disorders are associated with defects in social response and communication that often occur in the context of intellectual disability. Rett syndrome is one example in which epilepsy, motor impairment, and motor disturbance may co-occur. Mutations in histone demethylases
Chieko Kishi-Itakura et al.
Methods in molecular biology (Clifton, N.J.), 1759, 29-39 (2017-04-01)
In this chapter we describe the use of correlative light-electron microscopy (CLEM) to study, in cultured cells, the turnover of damaged mitochondria by PINK1/Parkin-dependent mitophagy. CLEM combines the advantages of light microscopy, which allows to image and rapidly screen a
Jiong Li et al.
Nature communications, 8, 15146-15146 (2017-04-26)
Human colorectal cancer stem cells (CSCs) are tumour initiating cells that can self-renew and are highly tumorigenic and chemoresistant. While genetic mutations associated with human colorectal cancer development are well-known, little is known about how and whether epigenetic factors specifically
Dan Luo et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(15), E3071-E3080 (2017-03-30)
The myogenic regulatory factor MyoD has been implicated as a key regulator of myogenesis, and yet there is little information regarding its upstream regulators. We found that Deltex2 inhibits myogenic differentiation in vitro, and that skeletal muscle stem cells from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service