Skip to Content
Merck
All Photos(1)

Key Documents

07-886

Sigma-Aldrich

Anti-phospho-PKR (Thr451) Antibody

Upstate®, from rabbit

Synonym(s):

EIF2AK2 (eukaryotic translation initiation factor 2-alpha kinase 2), EIF2AK1, MGC126524, PKR, PRKR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr451)

Gene Information

human ... EIF2AK2(5610)
mouse ... Eif2Ak2(19106)

Specificity

Recognizes phosphorylated PKR (Thr451)

Immunogen

Peptide corresponding to amino acid region encompassing the human phospho-PKR (Thr451)

Application

Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-phospho-PKR (Thr451) Antibody is validated for use in WB for the detection of phospho-PKR (Thr451).

Quality

Routinely evaluated by immunoblot.

Target description

68 kDa

Physical form

100 µL of affinity purified rabbit polyclonal IgG in Dulbecco’s PBS, containing 1.0 mg/mL BSA, 0.05% sodium azide and 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visnu Chaparro et al.
PLoS pathogens, 16(6), e1008291-e1008291 (2020-06-02)
The protozoan parasite Leishmania donovani (L. donovani) causes visceral leishmaniasis, a chronic infection which is fatal when untreated. Herein, we investigated whether in addition to altering transcription, L. donovani modulates host mRNA translation to establish a successful infection. Polysome-profiling revealed
P R Romano et al.
Molecular and cellular biology, 18(4), 2282-2297 (1998-04-07)
The human double-stranded RNA-dependent protein kinase (PKR) is an important component of the interferon response to virus infection. The activation of PKR is accompanied by autophosphorylation at multiple sites, including one in the N-terminal regulatory region (Thr-258) that is required
XuSha Zhou et al.
Viruses, 13(9) (2021-09-29)
Oncolytic virus (OV) as a promising therapeutic agent can selectively infect and kill tumor cells with naturally inherited or engineered properties. Considering the limitations of OVs monotherapy, combination therapy has been widely explored. MEK inhibitor (MEKi) Trametinib is an FDA-approved
Matthew J Elder et al.
European journal of immunology, 46(2), 455-463 (2015-11-18)
Thymic stromal lymphopoietin (TSLP) produced by epithelial cells acts on dendritic cells (DCs) to drive differentiation of TH 2-cells, and is therefore important in allergic disease pathogenesis. However, DCs themselves make significant amounts of TSLP in response to microbial products
Wei Li et al.
Scientific reports, 8(1), 166-166 (2018-01-11)
Cytoplasmic membrane-bound connexin 43 (Cx43) proteins oligomerize into hexameric channels (hemichannels) that can sometimes dock with hemichannels on adjacent cells to form gap junctional (GJ) channels. However, the possible role of Cx43 hemichannels in sterile and infectious inflammatory diseases has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service