Skip to Content
Merck
All Photos(1)

Documents

07-741

Sigma-Aldrich

Anti-AS160 (Rab-GAP) Antibody

serum, Upstate®

Synonym(s):

Acrg embryonic lethality minimal region ortholog, Akt substrate of 160 kDa, TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family, member

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

canine, human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TBC1D4(9882)
mouse ... Tbc1D4(210789)

General description

AS160 (Akt Substrate of 160 kDa, Akt1S1), Rab GAP (GTPase activating protein), is phosphorylated on multiple sites by Akt in response to insulin in adipocytes and contraction in muscles. AS160 is the long sought after link between the upstream insulin signaling through insulin/PI3K/Akt and GLUT4 vessicle translocation to the cell membrane. It has 6 Akt phosphorylation consensus sequences that are believed to become phosphorylated upon insulin stimulation. This phosphorylation is required for translocation of GLUT4-containing vesicles to the cell membrane1. A mutation of two or more of these phosphorylation sites results in a dramatic decrease in AS160 activity and insulin-stimulated GLUT4 redistribution at the cell membrane2. AS160 has also shown to be important for GLUT4 expcytosis2.

Specificity

Predicted to cross react with rat and dog based on sequence homology.
Recognizes AS160 (Rab-GAP), Mr 160 kDa.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 1178-1189 (C-PTNDKAKAGNKP) of rat Akt substrate of 160kDa (AS160) (Rab-GAP) with a N-terminal cysteine for conjugation purposes.

Application

Detect AS160 (Rab-GAP) using this Anti-AS160 (Rab-GAP) Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Routinely evaluated by Western Blot in RIPA lysates from 3T3/L1 cells differentiated into adipocytes.

Western Blot Analysis:
A 1:1000-1:5000 dilution of this lot detected AS160 (Rab-GAP) in RIPA lysates from 3T3/L1 cells differentiated into adipocytes by MDI treatment. This lot also detects AS160 (Rab-GAP) in HEK293 lysates.

Target description

Approx. 160 kDa

Physical form

Rabbit polyclonal antiserum IgG in buffer containing 0.05% sodium azide and 30% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
MDI-treated 3T3/L1 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clustering of GLUT4, TUG, and RUVBL2 protein levels correlate with myosin heavy chain isoform pattern in skeletal muscles, but AS160 and TBC1D1 levels do not.
Castorena, CM; Mackrell, JG; Bogan, JS; Kanzaki, M; Cartee, GD
Journal of Applied Physiology (1985)
AS160 phosphorylation is associated with activation of alpha2beta2gamma1- but not alpha2beta2gamma3-AMPK trimeric complex in skeletal muscle during exercise in humans.
Treebak, JT; Birk, JB; Rose, AJ; Kiens, B; Richter, EA; Wojtaszewski, JF
American Journal of Physiology. Endocrinology and Metabolism null
Joanne Bruno et al.
The Journal of cell biology, 214(1), 61-76 (2016-06-30)
RAB10 is a regulator of insulin-stimulated translocation of the GLUT4 glucose transporter to the plasma membrane (PM) of adipocytes, which is essential for whole-body glucose homeostasis. We establish SEC16A as a novel RAB10 effector in this process. Colocalization of SEC16A
TBC 11251. IPI 1040.
Drugs in R&D, 2(1), 38-39 (1999-12-28)
Myricetin Ameliorates Defective Post-Receptor Insulin Signaling via ?-Endorphin Signaling in the Skeletal Muscles of Fructose-Fed Rats.
Tzeng, TF; Liou, SS; Liu, IM
Evidence-Based Complementary and Alternative Medicine : ECAM null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service