Skip to Content
Merck
All Photos(2)

Key Documents

07-453

Sigma-Aldrich

Anti-B-Raf Antibody

Upstate®, from rabbit

Synonym(s):

Anti-B-RAF1, Anti-B-raf, Anti-BRAF-1, Anti-BRAF1, Anti-NS7, Anti-RAFB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BRAF(673)
mouse ... Braf(109880)
rat ... Rasd1(64455)

Specificity

full length recombinant BRaf (plus 6His tag) and B-Raf splice variants in cell lysates

Immunogen

synthetic peptide corresponding to amino acids 754-765 of human B-Raf

Application

Research Category
Signaling
Research Sub Category
MAP Kinases
This Anti-B-Raf Antibody is validated for use in IP, WB for the detection of B-Raf. This B-Raf antibody has been tested in Human, Mouse, and Rat.

Quality

routinely evaluated by immunoblot in RIPA lysates from PC12 cells and the full length purified B-Raf, active enzyme

Target description

94kDa

Linkage

Replaces: 04-328

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl,0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Ren et al.
International journal of cell biology, 2012, 913242-913242 (2012-05-23)
Recent high-throughput-sequencing of the cancer genome has identified oncogenic mutations in BRaf genetic locus as one of the critical events in melanomagenesis. In normal cells, the activity of BRaf is tightly regulated. Gain-of-function mutations like those identified in melanoma frequently
Sandeep S Joshi et al.
PloS one, 13(2), e0191264-e0191264 (2018-02-27)
Heat shock protein 90 (HSP90) is a molecular chaperone which stabilizes client proteins with important roles in tumor growth. 17-allylamino-17-demethoxygeldanamycin (17-AAG), an inhibitor of HSP90 ATPase activity, occupies the ATP binding site of HSP90 causing a conformational change which destabilizes
Lack of association between BRAF V600E mutation and mitogen-activated protein kinase activation in papillary thyroid carcinoma.
Hui Zuo,Yasushi Nakamura,Hironao Yasuoka,Ping Zhang,Misa Nakamura,Ichiro Mori et al.
Pathology International null
Regulation of B-Raf kinase activity by tuberin and Rheb is mammalian target of rapamycin (mTOR)-independent.
Karbowniczek, M; Cash, T; Cheung, M; Robertson, GP; Astrinidis, A; Henske, EP
The Journal of Biological Chemistry null
Kiril Bidzhekov et al.
Journal of cellular and molecular medicine, 11(6), 1395-1407 (2008-01-22)
Mouse embryonic endothelial progenitor cells (eEPCs) acquire a mature phenotype after treatment with cyclic adenosine monophosphate (cAMP), suggesting an involvement of Raf serine/threonine kinases in the differentiation process. To test this idea, we investigated the role of B-Raf and C-Raf

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service