Skip to Content
Merck
All Photos(1)

Key Documents

07-373

Sigma-Aldrich

Anti-acetyl-Histone H2B Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys5,Lys12,Lys15,Lys20)

Gene Information

human ... H2BC1(255626)

Specificity

acetylated Histone H2B

Immunogen

Synthetic peptide in which there is an acetyl-lysine at residues 5, 12, 15 and 20 of human Histone H2B

Application

Detect acetyl-Histone H2B also known as Histone H2B with Anti-acetyl-Histone H2B Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot in sodium butyrate treated, acid extracted proteins from HeLa cells

Target description

15 kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatial memory consolidation is associated with induction of several lysine-acetyltransferase (histone acetyltransferase) expression levels and H2B/H4 acetylation-dependent transcriptional events in the rat hippocampus.
Bousiges, O; Vasconcelos, AP; Neidl, R; Cosquer, B; Herbeaux, K; Panteleeva, I; Loeffler et al.
Neuropsychopharmacology null
Kelly L West et al.
Immunity, 23(2), 203-212 (2005-08-23)
V(D)J recombination is a tightly controlled process of somatic recombination whose regulation is mediated in part by chromatin structure. Here, we report that RAG2 binds directly to the core histone proteins. The interaction with histones is observed in developing lymphocytes
Estelle Schueller et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 33, 101-116 (2020-02-15)
Memory impairment is the main feature of Alzheimer's disease (AD). Initial impairments originate in the temporal lobe area and propagate throughout the brain in a sequential manner. Epigenetic mechanisms, especially histone acetylation, regulate plasticity and memory processes. These may be
Yixin Dong et al.
Molecular and cellular biology, 37(19) (2017-07-12)
Global histone hyperacetylation is suggested to play a critical role for replacement of histones by transition proteins and protamines to compact the genome during spermiogenesis. However, the underlying mechanisms for hyperacetylation-mediated histone replacement remains poorly understood. Here, we report that
A novel activator of CBP/p300 acetyltransferases promotes neurogenesis and extends memory duration in adult mice.
Chatterjee, S; Mizar, P; Cassel, R; Neidl, R; Selvi, BR; Mohankrishna, DV; Vedamurthy et al.
The Journal of Neuroscience null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service