Skip to Content
Merck
All Photos(1)

Key Documents

07-344

Sigma-Aldrich

Anti-CPI-17 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, pig, mouse, human, rabbit, bovine

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

CPI-17

Immunogen

Recombinant protein, full length pig CPI-17

Application

Anti-CPI-17 Antibody is an antibody against CPI-17 for use in WB.
NOT cross-reactive in yeast.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on RIPA lysates from mouse brain nuclear fraction and pig carotid artery tissue extract

Target description

22kDa

Physical form

Format: Purified
PBS, 0.02% sodium azide before the addition of glycerol to 10%
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution NMR structure of the myosin phosphatase inhibitor protein CPI-17 shows phosphorylation-induced conformational changes responsible for activation.
Ohki, S, et al.
Journal of Molecular Biology, 314, 839-849 (2001)
Sara R Turner et al.
Scientific reports, 9(1), 481-481 (2019-01-27)
The role of the smoothelin-like 1 (SMTNL1) protein in mediating vascular smooth muscle contractile responses to intraluminal pressure was examined in resistance vessels. Mesenteric arterioles from wild type (WT) and SMTNL1 global knock-out (KO) mice were examined with pressure myography.
M Eto et al.
The Journal of biological chemistry, 276(31), 29072-29078 (2001-06-09)
Histamine stimulus triggers inhibition of myosin phosphatase-enhanced phosphorylation of myosin and contraction of vascular smooth muscle. In response to histamine stimulation of intact femoral artery, a smooth muscle-specific protein called CPI-17 (for protein kinase C-potentiated inhibitory protein for heterotrimeric myosin
Cerebellar long-term synaptic depression requires PKC-mediated activation of CPI-17, a myosin/moesin phosphatase inhibitor
Eto, M., et al
Neuron, 36, 1145-1158 (2002)
Ayça Toprak Semiz et al.
Life sciences, 255, 117834-117834 (2020-05-27)
Hydrogen sulfide (H2S) is shown in ocular tissues and suggested to involve in the regulation of retinal circulation. However, the mechanism of H2S-induced relaxation on retinal artery is not clarified yet. Herein, we aimed to evaluate the role of several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service