Skip to Content
Merck

07-239

Anti-mGluR7 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-GLUR7, Anti-MGLU7, Anti-MGLUR7, Anti-NEDSHBA, Anti-PPP1R87

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
19
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRM7(2917)

General description

Metabotropic glutamate receptor 7 (UniProt: Q14831; also known as mGluR7) is encoded by the GRM7 (also known as GPRC1G, MGLUR7) gene (Gene ID: 2917) in human. GRM7 gene is mapped to human chromosome 3p26. L-glutamate is the major excitatory neurotransmitter in the central nervous system, and it activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into three groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. GRM7 belongs to Group III, and members of group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. mGluR7 is a multi-pass membrane protein and is synthesized with a signal peptide (aa 1-34), which is subsequently cleaved off to produce the mature protein (aa 35-915). mGluR7 is expressed in several parts of the brain, especially in the cerebral cortex, hippocampus, and cerebellum. It is located in the presynaptic zone of the synaptic cleft of glutamatergic synapses. Five isoforms of mGluR7 have been described that are produced by alternative splicing. The gene coding MGLUR7 is associated with autism and also participates in schizophrenia and bipolar disorder. Dysregulation of mGluR7 has been implicated in several neuropsychiatric disorders, including anxiety, depression, and fragile X syndrome (FXS), where its modulation by allosteric agents like AMN082 has shown therapeutic potential. Studies suggest that drugs targeting mGluR7: mGluR7 agonists, antagonists, and allosteric modulators may play a role as promising agents for the treatment of central nervous system disorders. (Ref.: Li P., et al. (2024). Ageing Res Rev. 102;102554; Palazzo E., et al. (2016). Curr Neuropharmacol. 14(5); 504-513).
Target molecular weight ~ 97 kDa observed; 102.25 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal cytoplasmic domain of human mGluR7.

Application

Anti-mGluR7, Cat. No. 07-239, is a rabbit polyclonal antibody that detects mGluR7 and is tested for use in Western Blotting and Dot Blot.
Research Category
Neuroscience
Tested Applications

Dot Blot Analysis: A 1:2,000 dilution from a representative lot detected synthetic mGluR7.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

This polyclonal antibody specifically detects mGluR7. It targets an epitope within 14 amino acids from the C-terminal cytoplasmic domain.

Physical form

Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide with 30% glycerol.

Preparation Note

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in rat brain microsomal preparation.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected mGluR7 in rat brain microsomal preparation.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Glutamate is an excitatory neurotransmitter found in the synaptic vesicles of glutamatergic synapses. The post-synaptic neurons in these synapses contain ionotropic and metabotropic glutamate receptors. Glutamate binds to AMPA (α-amino-3-hydroxy-5- methylisoxazole-4-propionic acid) subtype glutamate receptors, leading to sodium influx into the post-synaptic cell and resulting in neuronal excitability and synaptic transmission. The NMDA (N-methyl-d-aspartate) subtype glutamate receptors, on the other hand, regulate synaptic plasticity, and can influence learning and memory. The metabotropic g-protein coupled mGluRs modulate downstream calcium signaling pathways and indirectly influence the synapse’s excitability. The synaptic architecture includes intracellular scaffolding proteins (PSD-95, GRIP), intercellular cell adhesion molecules (NCAMs, N-Cadherins), and a variety of signaling proteins (CaMKII/PKA, PP1/PP2B). Processes critical for synaptic transmission and plasticity are influenced by these molecules and their interactions. When the function of these molecules is disrupted, it leads to synaptic dysfunction and degeneration, and can contribute to dementia as seen in Alzheimer’s disease.


Nicole M Fisher et al.
JCI insight, 6(4) (2021-01-22)
The metabotropic glutamate receptor 7 (mGlu7) is a G protein-coupled receptor that has been recently linked to neurodevelopmental disorders. This association is supported by the identification of GRM7 variants in patients with autism spectrum disorder, attention deficit hyperactivity disorder, and
Anatomy and function of group III metabotropic glutamate receptors in gastric vagal pathways.
Richard L Young,Nicole J Cooper,L Ashley Blackshaw
Neuropharmacology null
Tevye Jason Stachniak et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(39), 8150-8162 (2021-08-13)
Throughout development, neuronal identity is controlled by key transcription factors that determine the unique properties of a cell. During embryogenesis, the transcription factor Prox1 regulates VIP-positive cortical interneuron migration, survival, and connectivity. Here, we explore the role of Prox1 as



Global Trade Item Number

SKUGTIN
07-23904053252270352