Skip to Content
Merck
All Photos(2)

Key Documents

07-049

Sigma-Aldrich

Anti-Nedd4

Upstate®, from rabbit

Synonym(s):

E3 ubiquitin-protein ligase NEDD4, EC: 2.3.2.26, Cell proliferation-inducing gene 53 protein, HECT-type E3 ubiquitin transferase NEDD4, Neural precursor cell expressed developmentally down-regulated protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NDFIP1(80762)

General description

E3 ubiquitin-protein ligase NEDD4 (UniProt: P46934; also known as EC: 2.3.2.26, Cell proliferation-inducing gene 53 protein, HECT-type E3 ubiquitin transferase Nedd4, Neural precursor cell expressed developmentally down-regulated protein 4, NEDD-4) is encoded by the Nedd4 (also known as KIAA0093, Nedd4-1) gene (Gene ID: 4734) in human. NEDD4 is a peripheral membrane protein that is recruited to the plasma membrane by GRB10. Once complexed with GRB10 and IGF1R, it follows IGF1R internalization and remains associated with early endosomes. Nedd4 can accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. It is shown to specifically ubiquitinate Lysine 63 in target proteins. It also monoubiquitinates IGF1R at multiple sites, thus leading to receptor internalization and degradation in lysosomes. NEDD4 is reported to polyubiquitinate the epithelial sodium channel and any mutations that disrupt association between Nedd4 and sodium channel can lead to Liddle s syndrome, a hyperaldosteronism-like hypertension disease. NEDD4 can interact with viral proteins that contain a late-budding motif P-P-P-Y and this interaction is believed to be essential for viral particle budding of many retroviruses, like HTLV-1 Gag, and MLV Gag. It contains four WW domains (WW 1-4; aa 610-643; 767-800; 840-873; and 892-925) and one HECT domain (aa 984-1318). The WW domains mediate its interaction with PPxY motif-containing proteins.

Specificity

This rabbit polyclonal antibody specifically detects E3 ubiquitin-protein ligase Nedd4.

Immunogen

GST/His-tagged recombinant fragment corresponding to 68 amino acids from the N-terminal half of human Nedd4.

Application

Anti-Nedd4 Antibody, Cat. No. 07-049, is a rabbit polyclonal antibody that detects Nedd4 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative detected NEDD44 in rat cerebral cortex, rat cerebellum, and human colon cancer tissue sections.

Quality

Evaluated by Western Blotting in rat kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Nedd4 in rat kidney tissue lysate.

Target description

~110 kDa observed; 102.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 0.05% sodium azide.

Analysis Note

Control
Kidney cytoplasmic extracts.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinase-independent feedback of the TAK1/TAB1 complex on BCL10 turnover and NF-?B activation.
Moreno-Garcia, ME; Sommer, K; Rincon-Arano, H; Brault, M; Ninomiya-Tsuji, J; Matesic et al.
Molecular and cellular biology null
Nicola Amodio et al.
The American journal of pathology, 177(5), 2622-2634 (2010-10-05)
Loss of the PTEN tumor suppressor gene occurs frequently in non-small-cell lung carcinoma (NSCLC), although neither genetic alterations nor epigenetic silencing are significant predictors of PTEN protein levels. Since recent reports implicated neural precursor cell expressed, developmentally down-regulated 4-1 (NEDD4-1)
Efficiency of RNA interference in the mouse hematopoietic system varies between cell types and developmental stages.
Oberdoerffer, P; Kanellopoulou, C; Heissmeyer, V; Paeper, C; Borowski, C; Aifantis et al.
Molecular and cellular biology null
Taoling Zeng et al.
Cell reports, 7(3), 871-882 (2014-04-22)
RAS genes are among the most frequently mutated proto-oncogenes in cancer. However, how Ras stability is regulated remains largely unknown. Here, we report a regulatory loop involving the E3 ligase Nedd4-1, Ras, and PTEN. We found that Ras signaling stimulates
NEDD4-1 is a proto-oncogenic ubiquitin ligase for PTEN.
Wang, X; Trotman, LC; Koppie, T; Alimonti, A; Chen, Z; Gao, Z; Wang, J; Erdjument-Bromage et al.
Cell null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service