Skip to Content
Merck
All Photos(2)

Key Documents

06-138

Sigma-Aldrich

Anti-Cyclin A Antibody

Upstate®, from rabbit

Synonym(s):

Anti-CCN1, Anti-CCNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, sheep, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CCNA2(890)

General description

The cell cycle is regulated by the interplay of many molecules. Key among these are the cyclins which are expressed and then degraded in a concerted fashion to drive the stages of the cell cycle. Cyclins combine with cyclin dependent kinases (cdks) to form activated kinases that phosphorylate targets leading to cell cycle regulation. A breakdown in the regulation of this cycle can lead to out of control growth and contribute to tumor formation. Defects in many of the molecules that regulate the cell cycle have been implicated in cancer. Cyclin A is a member of the Cyclin A family that can bind and activate Cdc2 and Cdk2 and is involved in the control of mitosis and S phase. During S phase, cyclin A forms a complex with Cdk2, the pRB-related protein p107 and E2F.

Specificity

p58cyclin A; does not cross-react with cyclin B or cyclin D2, or 3

Immunogen

peptide corresponding amino acid residues 417-432 (C-KHGVSLLNPPETL-N-L)

Application

Detect Cyclin A with Anti-Cyclin A Antibody (Rabbit Polyclonal Antibody), that has been shown to work in IP, WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on HeLa nuclear extract

Target description

58kDa

Physical form

Format: Purified
Lyophilized
Protein G Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constitutive overexpression of CDK2 inhibits neuronal differentiation of rat pheochromocytoma PC12 cells
Dobashi, Y., et al
The Journal of Biological Chemistry, 270, 23031-23037 (1995)
Culture in low levels of oxygen enhances in vitro proliferation potential of satellite cells from old skeletal muscles.
M V Chakravarthy, E E Spangenburg, F W Booth
Cellular and Molecular Life Sciences null
Characterization of the cytoplasmic proline-directed protein kinase in proliferative cells and tissues as a heterodimer comprised of p34cdc2 and p58cyclin A.
Hall, F L, et al.
The Journal of Biological Chemistry, 266, 17430-17440 (1991)
J M Trowbridge et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(19), 10132-10137 (1997-09-18)
We have found that ectopic expression of cyclin A increases hormone-dependent and hormone-independent transcriptional activation by the estrogen receptor in vivo in a number of cell lines, including HeLa cells, U-2 OS osteosarcoma cells and Hs 578Bst breast epithelial cells.
J M Estanyol et al.
The Journal of biological chemistry, 274(46), 33161-33165 (1999-11-07)
The cyclin-dependent kinase (CDK) inhibitor p21(Cip1) has a dual role in the regulation of the cell cycle; it is an activator of cyclin D1-CDK4 complexes and an inhibitor of cyclins E/A-CDK2 activity. By affinity chromatography with p21(Cip1)-Sepharose 4B columns, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service