Skip to Content
Merck
All Photos(1)

Key Documents

06-127

Sigma-Aldrich

Anti-PDGF Antibody, neutralizing

Upstate®, from goat

Synonym(s):

Anti-PDGF-A, Anti-PDGF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, pig, mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PDGFA(5154)

Immunogen

Purified human PDGF-AB; recognizes PDGF AA, AB and BB.

Application

Anti-PDGF Antibody, neutralizing detects level of PDGF & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Target description

14/16 kDa

Physical form

DEAE chromatography
Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Feakins et al.
Human pathology, 31(10), 1214-1222 (2000-11-09)
The development of normal breast tissue and the pathogenesis of various tumors are influenced by growth factor-mediated epithelial-stromal interactions. Similar interactions may occur in fibroepithelial breast tumors. We have studied the expression of platelet-derived growth factor (PDGF) and PDGF beta
Chronic treatment with interleukin-1 beta induces coronary intimal lesions and vasospastic responses in pigs in vivo. The role of platelet-derived growth factor
Shimokawa, H., et al
The Journal of Clinical Investigation, 97, 769-776 (1996)
Bilal Alashkar Alhamwe et al.
Cellular & molecular immunology, 21(8), 918-931 (2024-06-29)
Recent studies reveal a critical role of tumor cell-released extracellular vesicles (EVs) in pancreatic cancer (PC) progression. However, driver genes that direct EV function, the EV-recipient cells, and their cellular response to EV uptake remain to be identified. Therefore, we
Luis Eduardo Becker et al.
Transplant international : official journal of the European Society for Organ Transplantation, 28(5), 553-564 (2015-01-30)
Histological evaluations of renal allograft biopsies are essential for diagnosis, but still show a low predictive value for long-term allograft function. One limitation relies on the fact that the analysis is usually based on a single biopsy sample, and therefore
Elyse Di Marco et al.
Free radical biology & medicine, 97, 556-567 (2016-07-23)
Smooth muscle cell (SMC) proliferation and fibrosis contribute to the development of advanced atherosclerotic lesions. Oxidative stress caused by increased production or unphysiological location of reactive oxygen species (ROS) is a known major pathomechanism. However, in atherosclerosis, in particular under

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service