Skip to Content
Merck
All Photos(1)

Documents

05-675

Sigma-Aldrich

Anti-MBP Antibody, clone SKB3

clone SKB3, Upstate®, from mouse

Synonym(s):

Anti-C76307, Anti-Hmbpr, Anti-R75289, Anti-golli-mbp, Anti-jve, Anti-mld, Anti-shi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SKB3, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MBP(4155)
mouse ... Mbp(17196)
rat ... Mbp(24547)

Specificity

Recognizes MBP.

Immunogen

Peptide corresponding to the human myelin basic protein sequence containing amino acid Thr98

Application

Detect MBP using this Anti-MBP Antibody, clone SKB3 validated for use in WB.
Research Category
Signaling
Research Sub Category
MAP Kinases

Quality

routinely evaluated by immunoblot on RIPA lysates from mouse and rat brain tissue preparations

Target description

18-20 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.02M Phosphate Buffer, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA containing no preservatives.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Giannotta et al.
EMBO molecular medicine, 6(2), 239-258 (2014-01-01)
Muscular dystrophies are severe genetic diseases for which no efficacious therapies exist. Experimental clinical treatments include intra-arterial administration of vessel-associated stem cells, called mesoangioblasts (MABs). However, one of the limitations of this approach is the relatively low number of cells
Samantha K Barton et al.
Frontiers in neuroscience, 15, 705306-705306 (2021-09-21)
Pathological hallmarks of amyotrophic lateral sclerosis (ALS), including protein misfolding, are well established in oligodendrocytes. More recently, an RNA trafficking deficit of key myelin proteins has been suggested in oligodendrocytes in ALS but the extent to which this affects myelination
Bangfu Zhu et al.
International journal of molecular sciences, 17(11) (2016-11-24)
The guided migration of neural cells is essential for repair in the central nervous system (CNS). Oligodendrocyte progenitor cells (OPCs) will normally migrate towards an injury site to re-sheath demyelinated axons; however the mechanisms underlying this process are not well
Robert Sparks et al.
The Journal of biological chemistry, 294(41), 14896-14910 (2019-08-16)
The metabolic consequences and sequelae of obesity promote life-threatening morbidities. PKCδI is an important elicitor of inflammation and apoptosis in adipocytes. Here we report increased PKCδI activation via release of its catalytic domain concurrent with increased expression of proinflammatory cytokines
Yuki Miyamoto et al.
Journal of cell science, 120(Pt 24), 4355-4366 (2007-11-29)
Oligodendrocyte precursor cells (OPCs) differentiate into oligodendrocytes (OLs) in order to form myelin, which is required for the rapid propagation of action potentials in the vertebrate nervous system. In spite of the considerable clinical importance of myelination, little is known

Articles

Derivation and characterization of functional human neural stem cell derived oligodendrocyte progenitor cells (OPCs) that efficiently myelinate primary neurons in culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service