Skip to Content
Merck
All Photos(1)

Key Documents

05-631

Sigma-Aldrich

Anti-IKKγ Antibody

Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IKBKG(8517)

General description

IKKγ/IKK3 is also known as NEMO, FIP-3 and IKKAP-1

Specificity

IKKγ/IKK3

Immunogen

His-tagged, full-length human IKKgamma/IKK3

Application

Anti-IKKγ Antibody is an antibody against IKKγ for use in WB.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

routinely evaluated by immunoblot on RIPA lysates from Jurkat cells

Target description

48kDa

Physical form

Format: Purified
PBS, pH 7.4, containing 0.05% sodium azide
Protein G Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oncoprotein Suppression of Tumor Necrosis Factor-induced NF{kappa}B Activation Is Independent of Raf-controlled Pathways
Hanson, J. L., et al
The Journal of Biological Chemistry, 278, 34910-34917 (2003)
Masaaki Morikawa et al.
Microbiology (Reading, England), 152(Pt 9), 2801-2807 (2006-09-02)
The extracellular matrix produced by Bacillus subtilis B-1, an environmental strain that forms robust floating biofilms, was purified, and determined to be composed predominantly of gamma-polyglutamate (gamma-PGA), with a molecular mass of over 1,000 kDa. Both biofilm formation and gamma-PGA
Sandeep Rana et al.
Chemical biology & drug design, 96(2), 773-784 (2020-04-03)
The release of an active drug from the prodrug generates a pro-fragment that typically has no biological activity and could result in adverse effects. By combining two drugs, wherein each drug acts as a pro-fragment of the other drug will
Dheeraj Soni et al.
Cell death discovery, 4, 60-60 (2018-05-26)
Vascular endothelial cadherin (VE-cad) expression at endothelial adherens junctions (AJs) regulates vascular homeostasis. Here we show that endothelial A20 is required for VE-cad expression at AJs to maintain and repair the injured endothelial barrier. In endothelial cell (EC)-restricted Tnfaip3 (A20)
Maarten F de Jong et al.
Nature microbiology, 1(7), 16084-16084 (2016-08-31)
The linear ubiquitin chain assembly complex (LUBAC) is a multimeric E3 ligase that catalyses M1 or linear ubiquitination of activated immune receptor signalling complexes (RSCs). Mutations that disrupt linear ubiquitin assembly lead to complex disease pathologies including immunodeficiency and autoinflammation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service