Skip to Content
Merck
All Photos(1)

Key Documents

05-311

Sigma-Aldrich

Anti-GST Tag Antibody, clone DG122-2A7

clone DG122-2A7, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DG122-2A7, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Immunogen

GST dervied from the pGEX expression vector, linked to KLH.

Application

Detect GST Tag using this Anti-GST Tag Antibody, clone DG122-2A7 validated for use in IP & WB.

Quality

routinely evaluated on GST fusion protein.

Target description

28-30 kDa plus fusion partner

Physical form

Format: Purified

Analysis Note

Control
Cells expressing the GST-tag

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional protein microarrays: just how functional are they?
Janie S Merkel,Gregory A Michaud,Michael Salcius,Barry Schweitzer,Paul F Predki
Current Opinion in Biotechnology null
Romesh R Subramanian et al.
International journal of oncology, 41(5), 1855-1862 (2012-08-28)
Members of the Ras protooncogene family are mutated in approximately 75% of colon cancers. The Raf kinases (Raf-1, b-Raf and a-Raf) directly interact with
Sun-Hong Hwang et al.
The Journal of biological chemistry, 295(8), 2336-2347 (2020-01-22)
Cell growth is positively controlled by the phosphoinositide 3-kinase (PI3K)-target of rapamycin (TOR) signaling pathway under conditions of abundant growth factors and nutrients. To discover additional mechanisms that regulate cell growth, here we performed RNAi-based mosaic analyses in the Drosophila
Shoshana Klein et al.
Protein expression and purification, 41(1), 162-169 (2005-04-02)
PKB/Akt is a protein involved in control of apoptosis, proliferation and cellular metabolism, and it has been found to be activated in many cancers. Activation of PKB involves recruitment of the enzyme by its PH domain to the cell membrane
Valeryi K Lishko et al.
Blood, 104(3), 719-726 (2004-04-20)
Plasminogen (Pg) has been implicated in many biologic processes involving extracellular proteolysis. We investigated whether Pg, by virtue of its capacity to be deposited within the extracellular matrix, can serve as a ligand for cell surface integrins. We report here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service