Skip to Content
Merck
All Photos(4)

Key Documents

04-835

Sigma-Aldrich

Anti-dimethyl-Histone H3 (Lys79) Antibody, clone NL59, rabbit monoclonal

culture supernatant, clone NL59, Upstate®

Synonym(s):

H3K79me2, Histone H3 (di methyl K79)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

NL59, monoclonal

species reactivity

vertebrates, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

shipped in

dry ice

target post-translational modification

dimethylation (Lys79)

Gene Information

human ... H3F3B(3021)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Dimethylation of Lys79 on histone H3 is associated with active gene transcription and is a marker for euchromatin. Histone H3 is methylated on Lys79 by the histone methyltransferase Dot1L..

Specificity

Broad species cross-reactivity expected due to sequence homology
Recognizes dimethyl-histone H3 (Lys79).

Immunogen

Epitope: Lys79
KLH-conjugated synthetic peptide containing the sequence DFme2KTD corresponding to dimethyl-lysine at position 79 of histone H3. The immunizing sequence is identical in human, mouse, rat and yeast. A C-terminal cysteine was added to facilitate conjugation.

Application

Anti-dimethyl-Histone H3 (Lys79) Antibody, clone NL59 is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys79) also known as H3K79me2, Histone H3 (di methyl K79) & has been validated in ChIP, WB, DB, Mplex.
Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 4 µL of Negative Control Supernatant, or 4 µL of Anti-dimethyl-Histone H3 (Lys79) and the Magna ChIP A Kit (Cat. # 17-610). Successful immunoprecipitation of dimethyl-Histone H3 (Lys79) associated DNA fragments was verified by qPCR using Control Primers
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Chromatin Immunoprecipitation: A representative lot of this antibody has been shown by an independent laboratory to work in ChIP.

ChIP-Seq Analysis:
A representative lot of this antibody was used by an independent laboratory for ChIP-Seq. See Egelhofer, T.A., et al. (2011). See Easwaran, H., et al. (2012).See Suzuki, H., et al. (2011).

Dot Blotting: Specificity of a representative lot confirmed by the ability of a 1:2500 dilution of the antibody to recognize peptides corresponding to regions of histone H3 with various modifications

Dot Blot Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No 04-835-S, Anti-dimethyl Histone H3 (Lys79) Antibody, clone NL59 (1:500 dilution). Proteins were visualized using a Donkey anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot of acid extracted proteins from HeLa cells

Target description

17 kDa

Linkage

Replaces: 05-835

Physical form

Culture supernantant in 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cathy J Spangler et al.
Cell reports, 38(7), 110369-110369 (2022-02-17)
DOT1L methylates histone H3 lysine 79 during transcriptional elongation and is stimulated by ubiquitylation of histone H2B lysine 120 (H2BK120ub) in a classical trans-histone crosstalk pathway. Aberrant genomic localization of DOT1L is implicated in mixed lineage leukemia (MLL)-rearranged leukemias, an
DNA methylation and histone modifications modulate the ?1,3 galactosyltransferase ?3Gal-T5 native promoter in cancer cells.
Anna Caretti,Silvia M Sirchia,Silvia Tabano,Aida Zulueta,Fabio Dall'olio,Marco Trinchera
The International Journal of Biochemistry & Cell Biology null
Julia M Schulze et al.
Molecular cell, 35(5), 626-641 (2009-08-18)
To identify regulators involved in determining the differential pattern of H3K79 methylation by Dot1, we screened the entire yeast gene deletion collection by GPS for genes required for normal levels of H3K79 di- but not trimethylation. We identified the cell
Eliza Mari Kwesi-Maliepaard et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(34), 20706-20716 (2020-08-09)
Cytotoxic T cell differentiation is guided by epigenome adaptations, but how epigenetic mechanisms control lymphocyte development has not been well defined. Here we show that the histone methyltransferase DOT1L, which marks the nucleosome core on active genes, safeguards normal differentiation
Eliza Mari Kwesi-Maliepaard et al.
Frontiers in genetics, 13, 1032958-1032958 (2022-11-26)
Cutaneous T-cell lymphomas (CTCLs) are a subset of T-cell malignancies presenting in the skin. The treatment options for CTCL, in particular in advanced stages, are limited. One of the emerging therapies for CTCL is treatment with histone deacetylase (HDAC) inhibitors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service