Skip to Content
Merck

Skip To

V-044

Vitamin K2-(MK-7)

100 μg/mL in acetonitrile, certified reference material, Cerilliant®

Synonym(s):

(all-E)–2-(3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaenyl)-3-methyl-1,4-naphthalenedione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C46H64O2
CAS Number:
Molecular Weight:
649.00
UNSPSC Code:
12352205
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

mp

-48 °C

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−70°C

SMILES string

O=C1c2c(cccc2)C(=O)C(=C1C\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)/C)/C)/C)/C)C

InChI

1S/C46H64O2/c1-34(2)18-12-19-35(3)20-13-21-36(4)22-14-23-37(5)24-15-25-38(6)26-16-27-39(7)28-17-29-40(8)32-33-42-41(9)45(47)43-30-10-11-31-44(43)46(42)48/h10-11,18,20,22,24,26,28,30-32H,12-17,19,21,23,25,27,29,33H2,1-9H3/b35-20+,36-22+,37-24+,38-26+,39-28+,40-32+

InChI key

RAKQPZMEYJZGPI-LJWNYQGCSA-N

General description

Vitamin K2 (MK-7), also known as menaquinone-7, is a homolog of Vitamin K2. Reports suggest that supplementation of Vitamin K2 (MK-7) can improve bone strength and prevent bone loss.

Application

This Certified Spiking Solution® is suitable for use as starting material in the preparation of linearity standards, calibrators, or controls in LC-MS/MS vitamin K2 testing applications such as assessment of vitamin K2 deficiency and detection and quantification of vitamin K2 in food or nutritional products
Vitamin K2 (MK-7) may be used as a certified reference material (CRM) for the determination of the analyte in biological samples and fermented foods by high performance liquid chromatography methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
47774V-031PHR2363
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

application(s)

clinical testing

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

application(s)

clinical testing

application(s)

pharmaceutical small molecule

format

single component solution

format

neat

format

single component solution

format

-

form

liquid

form

-

form

liquid

form

solid

Quality Level

300

Quality Level

100

Quality Level

300

Quality Level

300

grade

certified reference material

grade

analytical standard

grade

certified reference material

grade

certified reference material, pharmaceutical secondary standard


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters