Skip to Content
Merck
All Photos(1)

Key Documents

V-002

Supelco

Verapamil hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N2O4·HCl
CAS Number:
Molecular Weight:
491.06
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(CCN(C)CCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC

InChI

1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H

InChI key

DOQPXTMNIUCOSY-UHFFFAOYSA-N

General description

Verapamil, a phenylalkylamine analog, is used to treat high blood pressure and angina. This analytical reference standard is applicable for use as starting material in calibrators or controls for verapamil testing methods by LC-MS/MS or GC-MS. A calcium channel blocker, verapamil is sold under trade names Calan® and Covera-HS®.

Application


  • Estrone Calibration Solutions: These solutions are used for calibration in analytical techniques such as HPLC and mass spectrometry, providing reliability in the measurement of estrone and other estrogens in pharmaceutical and biochemical research (Moody et al., 2023).

  • Biochemical Research Reagents Estrone: As a biochemical research reagent, estrone is used in studies investigating its pharmacokinetics, metabolism, and its impact on cellular mechanisms, contributing to a deeper understanding of its therapeutic potentials and side effects (Johansson et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Calan is a registered trademark of G.D. Searle & Co.
Covera-HS is a registered trademark of G.D. Searle, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Ban Ke et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(3), 437-446 (2013-01-30)
Through PET imaging, our laboratory has studied the dynamic biodistribution of (11)C-verapamil, a P-gp substrate, in the nonhuman primate Macaca nemestrina. To gain detailed insight into the kinetics of verapamil transport across the blood-brain barrier (BBB) and the blood-placental barrier
Masato Ishigami et al.
Biochimica et biophysica acta, 1831(4), 683-690 (2013-01-12)
Although human MDR1 and MDR3 share 86% similarity in their amino acid sequences and are predicted to share conserved domains for drug recognition, their physiological transport substrates are quite different: MDR1 transports xenobiotics and confers multidrug resistance, while MDR3 exports
Weidong Jia et al.
Cancer chemotherapy and pharmacology, 71(6), 1585-1590 (2013-04-17)
This study was designed to evaluate efficacy, toxicity, and adverse effects of combination of chemotherapy drugs and intraperitoneal perfusion of verapamil in the treatment of malignant ascites. Seventy-two patients with malignant ascites were divided into two study groups. Patients in
Matylda Resztak et al.
Acta poloniae pharmaceutica, 70(3), 395-401 (2013-06-14)
A stereospecific capillary zone electrophoresis (CZE) method was developed for the determination of verapamil (VER) and its main metabolite norverapamil (NOR) in human plasma. Optimal temperature, cyclodextrin selectors (CDs), pH of background electrolyte (BGE) and voltage were established to obtain
Jian-Min Shen et al.
Pharmacological research, 70(1), 102-115 (2013-02-05)
In this paper we give a method of integrated treatment for cancer and drug-induced complications in the process of cancer therapy through dual-drug delivery system (DDDS). Two hydrophilic drugs, doxorubicin (an antitumor drug) and verapamil (an antiangiocardiopathy drug) combined preliminarily

Protocols

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service