Skip to Content
Merck
All Photos(1)

Key Documents

T-068

Supelco

Tilidine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO2 · HCl
CAS Number:
Molecular Weight:
309.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CCOC(=O)[C@@]1(CCC=C[C@H]1N(C)C)c2ccccc2

InChI

1S/C17H23NO2.ClH/c1-4-20-16(19)17(14-10-6-5-7-11-14)13-9-8-12-15(17)18(2)3;/h5-8,10-12,15H,4,9,13H2,1-3H3;1H/t15-,17+;/m1./s1

InChI key

MUWDJVKYGSDUSH-KALLACGZSA-N

General description

Tilidine is a synthetic opioid analgesic used mainly in Germany, Switzerland and Belgium for treatment of moderate to severe pain, both acute and chronic. While tilidine is considered a moderately-potent opioid, its active metabolite nortilidine possesses analgesic effects roughly equivalent to that of morphine.

Application

  • Opioid Detection in Hair: Research demonstrated the application of liquid chromatography/tandem mass spectrometry for detecting opioids, including Tilidine, in hair samples from palliative care patients, offering insights into long-term drug use and compliance monitoring (Musshoff et al., 2007).

  • Drug Testing in Hair Analysis: An analytical technique using LC-ESI-MS/MS and MALDI-MS/MS imaging was utilized to distinguish between Tilidine intake and external contamination in segmental hair analysis, improving accuracy in drug testing scenarios (Poetzsch et al., 2015).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul J Fudala et al.
Drug and alcohol dependence, 83 Suppl 1, S40-S47 (2006-03-28)
Non-medical abuse of prescription opioid medications is not a new phenomenon, but such use has been increasing in recent years. Various methods have been used and continue to be developed in an effort to limit diversion and abuse of opioid
[Low-dose strong opioid (LDSO)--treatment of pain in osteoarthritis].
W W Bolten et al.
MMW Fortschritte der Medizin, 150(51-52), 41-41 (2009-01-22)
Barbara Grün et al.
British journal of clinical pharmacology, 74(5), 854-863 (2012-03-03)
The analgesic activity of tilidine is mediated by its active metabolite, nortilidine, which easily penetrates the blood-brain barrier and binds to the µ-opioid receptor as a potent agonist. Tilidine undergoes an extensive first-pass metabolism, which has been suggested to be
[Tilidine-Naloxone sustained-release tablets for chronic pain associated with musculoskeletal disease].
U Junker et al.
MMW Fortschritte der Medizin, 148(37), 45-45 (2006-10-14)
Canli Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(22), 2089-2094 (2011-06-21)
We report in this manuscript, the use of direct ammonium persulfate-enhanced chemiluminescence (CL) imaging, to monitor changes to measure serum salbutamol concentration in subjects of different haptoglobin (Hp) phenotypes at different dosing time. It was noted that CL generated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service